Merck
  • Human Blood-Circulating Basophils Capture HIV-1 and Mediate Viral trans-Infection of CD4+ T Cells.
Human Blood-Circulating Basophils Capture HIV-1 and Mediate Viral trans-Infection of CD4+ T Cells.
Journal of virology (2015-05-29)
Ai-Ping Jiang, Jin-Feng Jiang, Ming-Gao Guo, Yong-Mei Jin, Yu-Ye Li, Jian-Hua Wang
ABSTRACT
Cell-associated HIV-1 infection has been proposed to play a pivotal role in the spread of HIV-1 infection. Granulocytes are a category of white blood cells, comprising mainly basophils, neutrophils, and eosinophils, and participate in various inflammatory reactions and defense against pathogens. Here, we investigated the role of human blood granulocytes in the dissemination of HIV-1. These cells were found to express a variety of HIV-1 attachment factors (HAFs). Basophils expressed HAFs dendritic cell (DC)-specific intercellular adhesion molecule 3 (ICAM3)-grabbing nonintegrin (DC-SIGN), DC immunoreceptor (DCIR), heparan sulfate proteoglycan (HSPG), and α4β7 integrin and mediated the most efficient capture of HIV-1 on the cell surface. Neutrophils were found to express DCIR and demonstrated limited efficiency of viral capture. Eosinophils expressed α4β7 integrin but exhibited little or no virus-binding capacity. Intriguingly, following direct contact with CD4+ T cells, viruses harbored on the surface of basophils were transferred to T cells. The contact between basophils and CD4+ T cells and formation of infectious synapses appeared necessary for efficient HIV-1 spread. In HIV-1-infected individuals, the frequency of basophils remained fairly stable over the course of disease, regardless of CD4+ T depletion or the emergence of AIDS-associated opportunistic infections. Collectively, our results provide novel insights into the roles of granulocytes, particularly basophils, in HIV-1 dissemination. Thus, strategies designed to prevent basophil-mediated viral capture and transfer may be developed into a new form of therapy. Cell-associated HIV-1 infection has been proposed to play a pivotal role in the spread of HIV-1 infection. Here, we demonstrated that human blood-circulating granulocytes, particularly basophils, can capture HIV-1 and mediate viral trans-infection of CD4+ T cells. The expression of a variety of HIV-1 attachment factors, such as the C-type lectins, etc., facilitates viral capture and transfer. Intriguingly, the frequency of basophils in patients with different levels of CD4+ T counts remains fairly stable during the course of disease. Our results provide novel insights into the roles of granulocytes, particularly basophils, in HIV-1 dissemination. We suggest that strategies designed to prevent basophil-mediated viral capture and transfer may be a new direction for the development of anti-HIV therapy.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Anti-gp120 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)