Skip to Content
Merck
All Photos(3)

Key Documents

C8106

Sigma-Aldrich

Calcium chloride

Synonym(s):

Calcium chloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaCl2 · 2H2O
CAS Number:
Molecular Weight:
147.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
crystals

grade

for analytical purposes

Quality Level

Agency

USP/NF

form

crystals

mp

176 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

SMILES string

O.O.Cl[Ca]Cl

InChI

1S/Ca.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LLSDKQJKOVVTOJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Levi et al.
Journal of thrombosis and haemostasis : JTH, 12(9), 1428-1436 (2014-05-09)
Four-factor prothrombin complex concentrates (PCCs), which contain factor II, FVII, FIX, and FX, have shown the potential to reverse the anticoagulant effect of rivaroxaban in healthy volunteers. The purpose of this study was to determine whether a three-factor PCC, which contains
J Philippe et al.
International journal of obesity (2005), 39(2), 295-302 (2014-06-04)
A significant proportion of severe familial forms of obesity remain genetically elusive. Taking advantage of our unique cohort of multigenerational obese families, we aimed to assess the contribution of rare mutations in 29 common obesity-associated genes to familial obesity, and
Maheswari Jayakannan et al.
Journal of experimental botany, 66(7), 1865-1875 (2015-01-24)
The role of endogenous salicylic acid (SA) signalling cascades in plant responses to salt and oxidative stresses is unclear. Arabidopsis SA signalling mutants, namely npr1-5 (non-expresser of pathogenesis related gene1), which lacks NPR1-dependent SA signalling, and nudt7 (nudix hydrolase7), which
Mohsen Akbari et al.
Advanced functional materials, 24(26), 4060-4067 (2014-11-21)
The fabrication of cell-laden structures with anisotropic mechanical properties while having a precise control over the distribution of different cell types within the constructs is important for many tissue engineering applications. Automated textile technologies for making fabrics allow simultaneous control
Michael A Goren et al.
Nature communications, 5, 5115-5115 (2014-10-09)
Opsin, the rhodopsin apoprotein, was recently shown to be an ATP-independent flippase (or scramblase) that equilibrates phospholipids across photoreceptor disc membranes in mammalian retina, a process required for disc homoeostasis. Here we show that scrambling is a constitutive activity of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service