Merck
  • Two main metabolites of gentiopicroside detected in rat plasma by LC-TOF-MS following 2,4-dinitrophenylhydrazine derivatization.
Two main metabolites of gentiopicroside detected in rat plasma by LC-TOF-MS following 2,4-dinitrophenylhydrazine derivatization.
Journal of pharmaceutical and biomedical analysis (2015-01-06)
Zhigang Wang, Shuhan Tang, Yan Jin, Yan Zhang, Masao Hattori, Hailong Zhang, Ning Zhang
ABSTRACT
The metabolism of gentiopicroside in vivo was studied by LC/MS following 2,4-dinitrophenylhydrazine derivatization for the first time. The ionization efficiency of the major metabolites erythrocentaurin and gentiopicral were greatly enhanced by the new analytical method developed, and they were successfully detected in rat plasma after oral administration of gentiopicroside. Methyl 4-formylbenzoate was used as the internal standard to quantify erythrocentaurin and gentiopicral in rat plasma in negative mode by UPLC-TOF-MS. It was found that erythrocentaurin reached the maximum plasma concentration of 625.2±246.3 ng/mL at about 2 h and gentiopicral reached the maximum plasma concentration of 157.6±86.6 ng/mL at about 4 h after oral administration of gentiopicroside at a dose of 200 mg/kg. The metabolic pathway of gentiopicroside to erythrocentaurin and gentiopicral was proposed. The monoterpene compound gentiopicroside was found to be metabolized to dihydroisocoumarin in vivo which may be responsible for the pharmacological effect of gentiopicroside. The results may shed light on clinical efficacy of gentiopicroside and the new analytical method developed may assist in studies for the metabolism of other natural iridoids and secoiridoids in vivo.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methyl 4-formylbenzoate, 99%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Gentiopicroside, ≥98% (HPLC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
2,4-Dinitrophenylhydrazine, reagent grade, 97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)