SML1572
Gentiopicroside
≥98% (HPLC)
Synonym(s):
(3S,4R)-4-ethenyl-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4,6-dihydro-3H-pyrano[3,4-c]pyran-8-one, (5R,6S)-5-Ethenyl-6-(-D-glucopyranosyloxy)-5,6-dihydro-1H,3H-Pyrano[3,4-c]pyran-1-one, (5R-trans)-5-ethenyl-6-(-D-glucopyranosyloxy)-5,6-dihydro-1H,3H-pyrano[3,4-c]pyran-1-one, Gentiopicrin, NSC606402
About This Item
Quality Level
Assay
≥98% (HPLC)
form
powder
optical activity
[α]/D -185 to -205°, c = 1 in H2O
color
white to beige
solubility
H2O: 20 mg/mL, clear
storage temp.
−20°C
SMILES string
C=C[C@H]1[C@H](O[C@]2([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)OC=C3C(OCC=C31)=O
InChI
1S/C16H20O9/c1-2-7-8-3-4-22-14(21)9(8)6-23-15(7)25-16-13(20)12(19)11(18)10(5-17)24-16/h2-3,6-7,10-13,15-20H,1,4-5H2/t7-,10-,11-,12+,13-,15+,16+/m1/s1
InChI key
DUAGQYUORDTXOR-GPQRQXLASA-N
Looking for similar products? Visit Product Comparison Guide
General description
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service