Merck
  • Active matrix metalloprotease-9 is associated with the collagen capsule surrounding the Madurella mycetomatis grain in mycetoma.
Active matrix metalloprotease-9 is associated with the collagen capsule surrounding the Madurella mycetomatis grain in mycetoma.
PLoS neglected tropical diseases (2014-03-29)
Kirsten Geneugelijk, Wendy Kloezen, Ahmed H Fahal, Wendy W J van de Sande
ABSTRACT
Madurella mycetomatis is the main causative organism of eumycetoma, a persistent, progressive granulomatous infection. After subcutaneous inoculation M. mycetomatis organizes itself in grains inside a granuloma with excessive collagen accumulation surrounding it. This could be contributing to treatment failure towards currently used antifungal agents. Due to their pivotal role in tissue remodelling, matrix metalloproteinases-2 (MMP-2) and 9 (MMP-9) or tissue inhibitor of metalloproteinases (TIMP) might be involved in this process. Local MMP-2 and MMP-9 expression was assessed by immunohistochemistry while absolute serum levels of these enzymes were determined in mycetoma patients and healthy controls by performing ELISAs. The presence of active MMP was determined by gelatin zymography. We found that both MMP-2 and MMP-9 are expressed in the mycetoma lesion, but the absolute MMP-2, -9, and TIMP-1 serum levels did not significantly differ between patients and controls. However, active MMP-9 was found in sera of 36% of M. mycetomatis infected subjects, whereas this active form was absent in sera of controls (P<0.0001). MMP-2, MMP-9, and TIMP-1 polymorphisms in mycetoma patients and healthy controls were determined through PCR-RFLP or sequencing. A higher T allele frequency in TIMP-1 (+372) SNP was observed in male M. mycetomatis mycetoma patients compared to controls. The presence of active MMP-9 in mycetoma patients suggest that MMP-9 is activated or synthesized by inflammatory cells upon M. mycetomatis infection. Inhibiting MMP-9 activity with doxycycline could prevent collagen accumulation in mycetoma, which in its turn might make the fungus more accessible to antifungal agents.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Human TIMP-1 ELISA Kit, for cell and tissue lysates
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)