Merck
  • Assessment of acylation routes and structural characterisation by liquid chromatography/tandem mass spectrometry of semi-synthetic acyl ester analogues of lipophilic marine toxins.
Assessment of acylation routes and structural characterisation by liquid chromatography/tandem mass spectrometry of semi-synthetic acyl ester analogues of lipophilic marine toxins.
Rapid communications in mass spectrometry : RCM (2014-11-05)
Pablo de la Iglesia, Elena Fonollosa, Jorge Diogène
ABSTRACT
Esterification is one of the most important metabolic routes of lipophilic marine toxins in shellfish. In this work we assessed several chemical acylation reactions aimed at obtaining acyl ester analogues via partial synthesis from the free toxins. The procedures developed including sensitive and selective methods based on liquid chromatography/tandem mass spectrometry (LC/MS/MS) can be applied to obtain reference materials that may be used as analytical standards (internal/external) for method development and calibration, as well as to perform toxicological in vitro and in vivo studies. Acylation systems involved both anhydrous and non-anhydrous fatty acid or acid halides as a source of the acyl radical, and several catalysers of the reaction. A series of mass spectrometric experiments involving product ion scans and multiple reaction monitoring (MRM) were used to confirm the identity and to elucidate the fragmentation pathways of the synthesised products. Reaction yields regarding reaction time and temperature were examined at sub-nmol scale for the acylation system consisting of palmitic anhydride and 4-(dimethylamino)pyridine (DMAP) in anhydrous pyridine, showing the best conditions at 75 °C for 60 min, 75 °C for 120 min and 100 °C for 270 min for cyclic imines, azaspiracid-1 and pectenotoxin-2, respectively. The esterification approach was verified at a larger scale for the esterification of gymnodimine-A (GYM-A), which kept a good yield >90% for the synthesis of 10-O-palmitoyl-GYM-A. Acyl ester analogues of lipophilic marine toxins have been synthesised and their structure elucidated by LC/MS/MS. For acyl ester analogues identical to natural metabolites, the procedures developed have potential to be applied for the semi-synthesis of metabolites in a sustainable, scalable and controlled way, avoiding extensive and tedious isolation and purification procedures from naturally contaminated shellfish. For the semi-synthetic esters structurally different than those found in shellfish, they may have applicability as internal standards for accurate quantifications of natural metabolites present in complex matrices.
Product Description
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Supelco
Triethylamine, analytical standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1-Methyl-2-pyrrolidinone, analytical standard
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Palmitoyl chloride, 98%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, purum, ≥99% (GC)
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%