Skip to Content
Merck
All Photos(1)

Documents

16304

Sigma-Aldrich

Triethylamine

purum, ≥99% (GC)

Synonym(s):

N,N-Diethylethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
Beilstein:
605283
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

3.5 (vs air)

Quality Level

vapor pressure

51.75 mmHg ( 20 °C)

grade

purum

Assay

≥99% (GC)

form

liquid

autoignition temp.

593 °F

expl. lim.

8 %

impurities

≤0.003% non-volatile matter

refractive index

n20/D 1.401 (lit.)

bp

88.8 °C (lit.)

mp

−115 °C (lit.)

density

0.726 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Zhao et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 6(5), 843-848 (2000-05-29)
The vinylation of iodobenzene with methyl acrylate has been studied with several supported palladium catalysts in N-methylpyrrolidone in the presence of triethylamine and/or sodium carbonate. The reaction can be performed in air without any solubilizing or activating ligands. It was
Acid-base reactions in non-dissociating solvents. Acetic acid and triethylamine in carbon tetrachloride and chloroform.
Barrow GM and Yerger EA.
Journal of the American Chemical Society, 76(20), 5211-5216 (1954)
THE SPECIFIC PRECIPITATION OF ORTHOPHOSPHATE AND SOME BIOCHEMICAL APPLICATIONS.
Y SUGINO et al.
The Journal of biological chemistry, 239, 2360-2364 (1964-07-01)
The asymmetric intramolecular Heck reaction in natural product total synthesis.
Amy B Dounay et al.
Chemical reviews, 103(8), 2945-2964 (2003-08-14)
Kai Morino et al.
Scientific reports, 5, 8953-8953 (2015-05-21)
Well-trained clinicians may be able to provide diagnosis and prognosis from very short biomarker series using information and experience gained from previous patients. Although mathematical methods can potentially help clinicians to predict the progression of diseases, there is no method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service