Merck
  • Activation of human 5-hydroxytryptamine type 3 receptors via an allosteric transmembrane site.
Activation of human 5-hydroxytryptamine type 3 receptors via an allosteric transmembrane site.
Molecular pharmacology (2014-10-24)
Stuart J Lansdell, Chaitra Sathyaprakash, Anne Doward, Neil S Millar
ABSTRACT
In common with other members of the Cys-loop family of pentameric ligand-gated ion channels, 5-hydroxytryptamine type 3 receptors (5-HT3Rs) are activated by the binding of a neurotransmitter to an extracellular orthosteric site, located at the interface of two adjacent receptor subunits. In addition, a variety of compounds have been identified that modulate agonist-evoked responses of 5-HT3Rs, and other Cys-loop receptors, by binding to distinct allosteric sites. In this study, we examined the pharmacological effects of a group of monoterpene compounds on recombinant 5-HT3Rs expressed in Xenopus oocytes. Two phenolic monoterpenes (carvacrol and thymol) display allosteric agonist activity on human homomeric 5-HT3ARs (64 ± 7% and 80 ± 4% of the maximum response evoked by the endogenous orthosteric agonist 5-HT, respectively). In addition, at lower concentrations, where agonist effects are less apparent, carvacrol and thymol act as potentiators of responses evoked by submaximal concentrations of 5-HT. By contrast, carvacrol and thymol have no agonist or potentiating activity on the closely related mouse 5-HT3ARs. Using subunit chimeras containing regions of the human and mouse 5-HT3A subunits, and by use of site-directed mutagenesis, we have identified transmembrane amino acids that either abolish the agonist activity of carvacrol and thymol on human 5-HT3ARs or are able to confer this property on mouse 5-HT3ARs. By contrast, these mutations have no significant effect on orthosteric activation of 5-HT3ARs by 5-HT. We conclude that 5-HT3ARs can be activated by the binding of ligands to an allosteric transmembrane site, a conclusion that is supported by computer docking studies.
Sigma-Aldrich
Linalool, 97%
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Carvacrol, ≥98%, FCC, FG
Sigma-Aldrich
Carvacrol, 98%
Sigma-Aldrich
p-Cymene, ≥97%, FG
Sigma-Aldrich
DL-Menthol, ≥95%, FCC, FG
Sigma-Aldrich
p-Cymene, 99%
Sigma-Aldrich
(±)-Menthol, racemic, ≥98.0% (GC)
Supelco
p-Cymene, analytical standard
Supelco
Carvacrol, analytical standard
Supelco
Linalool, analytical standard
Sigma-Aldrich
Carvacrol, natural, 99%, FG
Menthol, European Pharmacopoeia (EP) Reference Standard
Propofol, European Pharmacopoeia (EP) Reference Standard
Supelco
DL-Menthol, analytical standard
USP
Menthol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Sigma-Aldrich
Menthol, 99%
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Sigma-Aldrich
Thymol, FCC, FG
Sigma-Aldrich
Citral, mixture of cis and trans, ≥96%, FG
Sigma-Aldrich
(−)−α-Terpineol, natural, ≥96%, FCC, FG
Sigma-Aldrich
α-Terpineol, 90%, technical grade
Sigma-Aldrich
Citral, natural, ≥96%, FCC, FG
Supelco
(−)-Linalool, analytical standard
Supelco
Thymol, analytical standard
Sigma-Aldrich
4-Isopropyl-3-methylphenol, 99%
Sigma-Aldrich
Citral, 95%
Sigma-Aldrich
Terpineol, mixture of isomers, 96%, FG
Sigma-Aldrich
Thymol, ≥98.5%