Skip to Content
Merck
All Photos(2)

Documents

C83007

Sigma-Aldrich

Citral

95%

Synonym(s):

Lemonal, 3,7-Dimethyl-2,6-octadienal, Geranial and neral mixture

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCHO
CAS Number:
Molecular Weight:
152.23
Beilstein:
1721871
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

5 (vs air)

vapor pressure

0.2 mmHg ( 200 °C)

Assay

95%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

229 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)C=C(C)CC\C=C(\C)C

InChI

1S/C10H16O/c1-9(2)5-4-6-10(3)7-8-11/h5,7-8H,4,6H2,1-3H3

InChI key

WTEVQBCEXWBHNA-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

General description

Citral is a mixture of neral (cis isomer of 3,7-dimethyl-2,6-octadienal) and geranial (trans isomer of 3,7-dimethyl-2,6-octadienal). It exhibits antimicrobial activity and pheromonal effects and is generally used for the synthesis of vitamin A and ionones.

Citral is a monoterpene aldehyde used as an aromatic ingredient in food and cosmetic industry.

Application

  • Citral has been used as an anti-tumor agent against breast cancer cell lines (MCF-7 and MDA-MB-231).
  • It shows antipyretic effects which can be studied by systemic administration of lipopolysaccharide (LPS) to rats.
  • It can undergo 1,3-dipolar cycloaddition reaction with trimethylsilylazide to form 1,2,3-triazolines which possess pharmaceutical properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - DIN 51758

Flash Point(C)

98 °C - DIN 51758

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theoretical and experimental studies of 1, 3-dipolar cycloaddition reactions between trimethylsilylazide and citral (geranial and neral)
Taban S and Taherpour AA
Indian Journal of Chemistry, 55(B), 1400-1414 (2016)
Clarissa M D Mota et al.
Journal of ethnopharmacology, 250, 112486-112486 (2019-12-18)
Citral (3,7-dimethyl-2,6-octadienal) is the main component of Cymbopogon citratus (DC) Stapf, an herb with analgesic properties. Arthritic pain is the main unpleasant component of rheumatoid arthritis. The pharmacological approaches used to treat arthritic pain are often accompanied by adjuvant drugs
Citral induced apoptosis in MDA-MB-231 spheroid cells
Nigjeh SE, et al.
BMC Complementary and Alternative Medicine, 18(1), 56-56 (2018)
Chemical and antimicrobial studies of monoterpene: Citral
Saddiq, Amna A and Khayyat, et al.
Pesticide Biochemistry and Physiology, 98, 89-93 (2010)
Antipyretic effects of citral and possible mechanisms of action
Emilio-Silva MT, et al.
Inflammation, 40(5), 1735-1741 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service