Skip to Content
Merck
All Photos(1)

Documents

W266507

Sigma-Aldrich

DL-Menthol

≥95%, FCC, FG

Synonym(s):

Menthol, 2-Isopropyl-5-methylcyclohexanol, Hexahydrothymol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
FEMA Number:
2665
Beilstein:
3194263
EC Number:
Council of Europe no.:
63
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.015
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor pressure

0.8 mmHg ( 20 °C)

Assay

≥95%

bp

216 °C (lit.)

mp

34-36 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

woody; minty; cooling

SMILES string

CC(C)C1CCC(C)CC1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3

InChI key

NOOLISFMXDJSKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-Menthol is a racemic mixture of D- and L-menthol. Its optical resolution produces L-menthol, which is widely used in confectionaries cosmetics and toothpaste for its characteristic peppermint flavor and cooling effect.

Application


  • Developing of an eco-friendly liquid-liquid microextraction method by using menthol-based hydrophobic deep eutectic solvent for determination of basic fuchsin dye: assessment of the greenness profile.: This study introduces a sustainable method using DL-menthol-based solvents for extracting and analyzing industrial dyes, providing a green alternative to traditional solvent extraction techniques (Aydin et al., 2024).

  • Potential fumigant toxicity of essential oils against Sitotroga cerealella (Olivier) (Lepidoptera: Gelechiidae) and its egg parasitoid Trichogramma evanescens (Hymenoptera: Trichogrammatidae).: This research highlights the efficacy of DL-menthol in pest management, examining its potential as a natural fumigant that could replace synthetic chemicals in agricultural practices (Elbehery et al., 2024).

  • Differential analysis of volatiles in five types of mosquito-repellent products by chemometrics combined with headspace GC-Orbitrap HRMS nontargeted detection.: Explores the application of DL-menthol in developing mosquito-repellent formulations, enhancing the understanding of volatile compounds in public health products (Zhao et al., 2024).

  • Vortex-assisted dispersive liquid-liquid microextraction based on the hydrophobic deep eutectic solvent-based ferrofluid for extraction and detection of myclobutanil.: Utilizes DL-menthol in innovative extraction methods for pesticide analysis, promoting safer and more efficient detection techniques in food safety and environmental monitoring (Fattahi et al., 2023).

Biochem/physiol Actions

Taste at 25 ppm

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

204.8 °F

Flash Point(C)

96 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucie Percevault et al.
The journal of physical chemistry. B, 124(41), 9126-9135 (2020-09-19)
We have performed small-angle neutron scattering in a momentum transfer range (0.05 < Q < 0.5 Å-1) to study long-range order and concentration fluctuations in deep eutectic solvents (DESs) and their aqueous solutions. Ethaline (choline chloride/ethylene glycol), glycerol/lactic acid, and
Optimized synthesis of lipase-catalyzed l-menthyl butyrate by Candia rugosa lipase.
Shih L, et al.
Food Chemistry, 100(3), 1223-1228 (2007)
Joanne D'Silva et al.
American journal of preventive medicine, 43(5 Suppl 3), S242-S248 (2012-10-25)
Menthol cigarettes account for 25% of the market in the U.S. The Food and Drug Administration currently is considering regulatory action on tobacco products, including a ban on menthol cigarettes. With 39% of menthol smokers reporting that they would quit
Miranda R Jones et al.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology, 22(2), 224-232 (2012-12-20)
In the United States, cigarette flavorings are banned, with the exception of menthol. The cooling effects of menthol could facilitate the absorption of tobacco toxicants. We examined levels of biomarkers of tobacco exposure among U.S. smokers of menthol and nonmenthol
V Blair Journigan et al.
Life sciences, 92(8-9), 425-437 (2012-11-20)
Since the discovery of the TRPM8 gene in 2001, the TRPM8 ion channel, better known as the 'cold receptor' has been the target of a significant effort from the pharmaceutical industry to produce small-molecule agonists and antagonists of this receptor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service