Merck
  • Effects of additives and templates on calcium carbonate mineralization in vitro.
Effects of additives and templates on calcium carbonate mineralization in vitro.
Micron (Oxford, England : 1993) (2010-10-19)
Dongni Ren, Qingling Feng, Xavier Bourrat
ABSTRACT
The review focuses on the effects of several important additives and templates controlling the calcium carbonate crystals formation and the complexity of the crystal morphologies in vitro. Additives include soluble matrices extracted from shells and pearls, amino-acids, magnesium ions and collagen among others. Templates include modified single crystal silicon, natural biominerals among others. Mechanisms proposed to explain the phenomena are not systematic, further studies are necessary to explain how organic matrices mediate calcium carbonate mineralization.
Product Description
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder