Skip to Content
Merck
All Photos(3)

Key Documents

202932

Sigma-Aldrich

Calcium carbonate

≥99.995% trace metals basis

Synonym(s):

Aragonite, Calcium monocarbonate, Chalk, Limestone, Monocalcium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.995% trace metals basis
form:
powder and chunks
solubility:
H2O: insoluble
Pricing and availability is not currently available.

Quality Level

Assay

≥99.995% trace metals basis

form

powder and chunks

impurities

≤50.0 ppm Trace Metal Analysis

solubility

H2O: insoluble

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium carbonate is an inorganic compound that is sparingly soluble in water but highly soluble in dilute acids. It plays a crucial role in material science, particularly in the development of solar cells, batteries, and supercapacitors. In solar cell, it acts as a precursor material for the perovskite layer, enhancing the stability and performance of perovskite solar cells. Additionally, calcium carbonate is utilized in electrode production, serving as a conductive additive in cathode or anode materials, thus improving overall battery performance and durability.

Application

Calcium carbonate can be used as:     
  • A template, graphitization catalyst, and activating agent in the green synthesis of porous graphitic carbons from coal tar pitch for high-performance lithium-ion batteries.     
  • A raw material to synthesize calcium titanate (CaTiO3) perovskite material through sintering with titanium oxide (TiO2) by the sol-gel method. Calcium titanate can be used to create large-scale, low-cost, and sustainable solar cells.
  • A catalyst supports for improving the performance and durability of fuel cells, thereby contributing to the advancement of clean energy technologies.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Azizur Rahman et al.
PloS one, 8(3), e58781-e58781 (2013-03-26)
An organic matrix consisting of a protein-polysaccharide complex is generally accepted as an important medium for the calcification process. While the role this "calcified organic matrix" plays in the calcification process has long been appreciated, the complex mixture of proteins
Jane A Cauley et al.
Journal of women's health (2002), 22(11), 915-929 (2013-10-18)
Clinical outcomes of the Women's Health Initiative (WHI) calcium plus vitamin D supplementation trial have been reported during 7.0 years of active intervention. We now report outcomes 4.9 years after the intervention stopped and cumulative findings. Postmenopausal women (N=36,282) were
Julyan H E Cartwright et al.
Angewandte Chemie (International ed. in English), 51(48), 11960-11970 (2012-11-06)
Although the polymorphism of calcium carbonate is well known, and its polymorphs--calcite, aragonite, and vaterite--have been highly studied in the context of biomineralization, polyamorphism is a much more recently discovered phenomenon, and the existence of more than one amorphous phase
Christopher T Parsons et al.
Environmental pollution (Barking, Essex : 1987), 178, 254-263 (2013-04-17)
Arsenic contamination of floodplain soils is extensive and additional fresh arsenic inputs to the pedosphere from human activities are ongoing. We investigate the cumulative effects of repetitive soil redox cycles, which occur naturally during flooding and draining, on a calcareous
Pedram Fatehi et al.
Carbohydrate polymers, 94(1), 531-538 (2013-04-03)
In this work, we aimed at adsorbing the oligo-sugars of prehydrolysis liquor on precipitated calcium carbonate (PCC) to produce modified PCC. The results showed that the adsorptions of oligo-sugars, lignin and furfural were greater on porous PCC (PCC2) than on

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service