Skip to Content
Merck
All Photos(1)

Documents

1086403

USP

Calcium Carbonate (AS)

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Calcium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

calcium carbonate

manufacturer/tradename

USP

density

2.93 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Calcium Carbonate (AS) USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polysaccharides and proteoglycans in calcium carbonate-based biomineralization.
José L Arias et al.
Chemical reviews, 108(11), 4475-4482 (2008-07-26)
Ling Li et al.
Nature materials, 13(5), 501-507 (2014-04-01)
Hierarchical composite materials design in biological exoskeletons achieves penetration resistance through a variety of energy-dissipating mechanisms while simultaneously balancing the need for damage localization to avoid compromising the mechanical integrity of the entire structure and to maintain multi-hit capability. Here
Claudia Gagnon et al.
PloS one, 9(10), e109607-e109607 (2014-10-10)
To examine whether combined vitamin D and calcium supplementation improves insulin sensitivity, insulin secretion, β-cell function, inflammation and metabolic markers. 6-month randomized, placebo-controlled trial. Ninety-five adults with serum 25-hydroxyvitamin D [25(OH)D] ≤55 nmol/L at risk of type 2 diabetes (with
Ruqian Qin et al.
PloS one, 9(7), e103477-e103477 (2014-07-30)
Trees in the genus Populus and their interspecific hybrids are used across North America for fiber production and as a potential source of biofuel. Plantations of these species are severely impacted by a fungal pathogen, Sphaerulina musiva, the cause of
Dongni Ren et al.
Micron (Oxford, England : 1993), 42(3), 228-245 (2010-10-19)
The review focuses on the effects of several important additives and templates controlling the calcium carbonate crystals formation and the complexity of the crystal morphologies in vitro. Additives include soluble matrices extracted from shells and pearls, amino-acids, magnesium ions and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service