Merck
  • Preparation and transdermal diffusion evaluation of the prazosin hydrochloride-loaded electrospun poly(vinyl alcohol) fiber mats.
Preparation and transdermal diffusion evaluation of the prazosin hydrochloride-loaded electrospun poly(vinyl alcohol) fiber mats.
Journal of nanoscience and nanotechnology (2014-04-25)
Xiaobing Shen, Qian Xu, Shi Xu, Jie Li, Niping Zhang, Ling Zhang
ABSTRACT
This study reports on the use of electrospun polyvinyl alcohol (PVA) nanofiber mats loaded with prazosin hydrochloride (PRH) as a transdermal drug delivery system, investigating the morphology of electrospun PVA nanofibers, the in vitro release characteristics of the drug from the as-spun fibers, and the influence of permeation enhancer (water-resoluble azone, WSA) on transdermal diffusion of PRH through a rat skin. The same was also conducted on the PRH -loaded as-cast PVA films for comparison. Results indicated that the morphology of PRH-loaded PVA fibers observed by scanning electron microscopy (SEM) relied on the electrospinning processing parameters, and the addition of WSA had obvious effects on the diameter and morphology of electrospun PVA fibers. The PRH-loaded electrospun PVA fiber mats exhibited much higher accumulated release dose and release rate of PRH than as-cast PVA films. And WAS can improve the release amount and rate of PRH from drug-loaded samples. The content of PRH in receiver was more than that in the stratum corneum and in the dermis. It was concluded that the PRH-loaded electropun PVA fiber mats as a transdermal patches can be a promising candidate for the conventional preparation.
Sigma-Aldrich
Prazosin hydrochloride, ≥99.0% (HPLC)
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
USP
Prazosin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Prazosin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
Poly(vinyl alcohol), European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed