Skip to Content
Merck
All Photos(2)

Key Documents

363103

Sigma-Aldrich

Poly(vinyl alcohol)

average Mw 146,000-186,000, 87-89% hydrolyzed

Synonym(s):

PVA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.23

form

powder or crystals

mol wt

average Mw 146,000-186,000

falling ball

40-50 cP, 4 % in H2O(20 °C)(lit.)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(vinyl alcohol) (PVA) is a vinyl based water soluble polymer that can be used as a non-ionic surface active agent. It can also be used as a biodegradable polymer and can be incorporated in adhesives, coatings, textiles, ceramics and cosmetics. Its properties include good transparency, chemical resistance, toughness and anti-electrostatic properties.

Application

PVA can be used in the preparation of hydrogel templates for drug delivery applications. It can also be used in the development of organic memory systems.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plastics Materials, 386-397 (1999)
Coexistence of Write Once Read Many Memory and Memristor in blend of Poly (3, 4-ethylenedioxythiophene): polystyrene sulfonate and Polyvinyl Alcohol
Nguyen VC and Lee PS
Scientific Reports, 6(1-2), 38816-38816 (2016)
Compostable Polymer Properties and Packaging Applications
Plastic Films in Food Packaging, 217-248 (2013)
Microparticles produced by the hydrogel template method for sustained drug delivery
Lu Y, et al.
International Journal of Pharmaceutics, 461(1-2), 258-269 (2014)
Biodegradation of poly (vinyl alcohol) based materials
Chiellini E, et al.
Progress in Polymer Science, 28(6), 963-1014 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service