Skip to Content
Merck
All Photos(1)

Key Documents

10853

Sigma-Aldrich

Mowiol® 6-98

Mw ~47,000

Synonym(s):

Poly(vinyl alcohol)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

Quality Level

mol wt

Mw ~47,000

extent of labeling

98.0-98.8 mol% hydrolysis
~1,000 polymerization

ign. residue

≤0.5%

viscosity

5-7 mPa.s, 4 % in H2O(20 °C)

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Mowiol is a registered trademark of Kuraray Europe GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelly E Coller et al.
PLoS pathogens, 5(12), e1000702-e1000702 (2009-12-31)
Hepatitis C virus (HCV) enters hepatocytes following a complex set of receptor interactions, culminating in internalization via clathrin-mediated endocytosis. However, aside from receptors, little is known about the cellular molecular requirements for infectious HCV entry. Therefore, we analyzed a siRNA
Sreejith Raveendran et al.
Carbohydrate polymers, 92(2), 1225-1233 (2013-02-13)
Extremophilic bacterial polysaccharide based biocompatible nanofibers were produced for the first time via electrospinning technique. Mauran (MR), an extremophilic sulfated exopolysaccharide was extracted from moderately halophilic bacterium, Halomonas maura and characterized for the application of nanofiber synthesis. Thin-uniform MR nanofibers
Feng-yong Liu et al.
Acta radiologica (Stockholm, Sweden : 1987), 54(1), 61-66 (2013-02-05)
Preoperative embolization of tumors is a well-established procedure that has been successfully applied in various clinical situations. Preoperative embolization can reduce the vascularity of tumors resulting in a clearer operative field, less difficult dissection, decreased blood loss, and, in some
Bor-Sen Chiou et al.
International journal of biological macromolecules, 55, 214-220 (2013-01-29)
Pollock gelatin/poly(vinyl alcohol) (PVA) fibers were electrospun using deionized water as the solvent and pollock gelatin/poly(lactic acid) (PLA) fibers were electrospun using 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) as the solvent. The chemical, thermal, and thermal stability properties were examined for the electrospun samples.
Cijun Shuai et al.
Biofabrication, 5(1), 015014-015014 (2013-02-07)
A tetragonal polyvinyl alcohol (PVA) scaffold with 3D orthogonal periodic porous architecture was fabricated via selective laser sintering (SLS) technology. The scaffold was fabricated under the laser power of 8 W, scan speed of 600 mm min(-1), laser spot diameter of 0.8 mm

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service