Skip to Content
Merck
All Photos(2)

Documents

P1763

Sigma-Aldrich

Poly(vinyl alcohol)

Fully hydrolyzed

Synonym(s):

PVA, PVOH, Poly(ethenol)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CHOH-]n
CAS Number:
MDL number:
UNSPSC Code:
12352104
NACRES:
NA.75

Quality Level

form

powder

color

colorless

viscosity

11-14 cP, 4 % in H2O(20 °C)(lit.)

solubility

H2O: soluble (hot)

density

1.080 g/cm3

application(s)

hematology
histology

storage temp.

room temp

InChI

1S/C2H4O/c1-2-3/h2-3H,1H2

InChI key

IMROMDMJAWUWLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyvinyl alcohol (PVOH) is a hydrophilic linear polymer that forms copolymers of vinyl alcohol and vinyl acetate. It is prepared by alkaline hydrolysis. It is known to possess high mechanical strength, biocompatibility, and non-toxicity. Modified- PVOH structures are useful in biomedical applications.

Application

Poly(vinyl alcohol) has been used in the preparation of multilayer active film for its antimicrobial analysis. It has also been used to study the effect of protein supplements on porcine parthenotes.
Poly(vinyl alcohol) is also suitable for:
  • in vitro production (IVP) of bovine embryos
  • oocyte recovery and in vitro maturation
  • as a macromolecular component in basal cultured medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Controlled release of antimicrobial compounds from highly swellable polymers.
Buonocore G G, et al.
Journal of Food Protection, 67(6), 1190-1194 (2004)
Rasha Al-Attar et al.
Cryobiology, 86, 10-18 (2019-01-15)
Wood frogs, Rana sylvatica, can undergo prolonged periods of whole body freezing during winter, locking as much as 65-70% of total body water into extracellular ice and imposing both anoxia and dehydration on their cells. Metabolic rate depression (MRD) is
Zinan He et al.
Medical engineering & physics, 77, 60-68 (2020-01-20)
Abdominal aortic aneurysm (AAA) is an asymptomatic condition due to the dilation of abdominal aorta along with progressive wall degeneration, where rupture of AAA is life-threatening. Failures of AAA endovascular repair (EVAR) reflect our inadequate knowledge about the complex interaction
Fetal bovine serum influences apoptosis and apoptosis-related gene expression in porcine parthenotes developing in vitro.
Cui X S, et al.
Reproduction (Cambridge, England), 127(1), 125-130 (2004)
Gang Wang et al.
PloS one, 6(11), e27605-e27605 (2011-11-24)
Mucosal vaccination has been demonstrated to be an effective means of eliciting protective immunity against aerosol infections of foot and mouth disease virus (FMDV) and various approaches have been used to improve mucosal response to this pathogen. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service