Merck
  • Diagnosis of tuberculous meningitis by detection of tuberculostearic acid in cerebrospinal fluid.
Diagnosis of tuberculous meningitis by detection of tuberculostearic acid in cerebrospinal fluid.
Lancet (London, England) (1987-07-18)
G L French, R Teoh, C Y Chan, M J Humphries, S W Cheung, G O'Mahony
ABSTRACT
Tuberculostearic acid, a structural component of Mycobacterium tuberculosis, was identified by gas chromatography/mass spectrometry with selected ion monitoring in cerebrospinal fluid (CSF) from 13 patients with proven and 8 out of 9 patients with suspected tuberculous meningitis; the negative result was in a patient whose symptoms and CSF abnormalities may have been due to systemic lupus erythematosus. Tuberculostearic acid was found in the CSF of only 1 patient out of 87 with non-tuberculous meningitis or non-infectious disorders; the single false-positive result was probably caused by intrathecal treatment with amikacin. Other aminoglycosides and antituberculous drugs did not interfere with the assay. In serial CSF samples from patients with tuberculous meningitis tuberculostearic acid was still present after 8 months of continuous supervised chemotherapy. Detection of tuberculostearic acid in CSF is a rapid, sensitive, and specific test for tuberculous meningitis, and can be used for retrospective diagnosis in patients who have been started on therapy.
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
2,3-Pyridinedicarboxylic acid, 99%
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pipecolinic acid, 98%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Millipore
Urea solution, suitable for microbiology, 40% in H2O