Skip to Content
Merck
All Photos(1)

Key Documents

A2129

Sigma-Aldrich

γ-Aminobutyric acid

≥99% (TLC), powder or crystals, inhibitory neurotransmitter

Synonym(s):

GABA, Gammalon, gamma-Aminobutyric acid, 3-Carboxypropylamine, 4-Aminobutanoic acid, Piperidic acid, Piperidinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3COOH
CAS Number:
Molecular Weight:
103.12
Beilstein:
906818
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

γ-Aminobutyric acid, ≥99%

Quality Level

Assay

≥99%

form

powder or crystals

storage condition

(Tightly closed. Dry.)

color

colorless

mp

195 °C (dec.) (lit.)

solubility

water: 50 mg/mL, clear, colorless

SMILES string

NCCCC(O)=O

InChI

1S/C4H9NO2/c5-3-1-2-4(6)7/h1-3,5H2,(H,6,7)

InChI key

BTCSSZJGUNDROE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

γ-Aminobutyric acid (GABA) is a non-amino protein. It is found in plants, animals, and microorganisms. GABA has been used in pharma and food industries as a component of cheese, shochu, gabaron tea, and gammalone.[1]
Research area: Neuroscience

Application

γ-Aminobutyric acid (GABA) has been used:
  • in the preparation of GABA–BSA (bovine serum albumin)–GA (glutaraldehyde) conjugate, which is used as a control for GABA immunostaining[2]
  • in the macromolecule suppressed detection of GABA by magnetic resonance spectroscopy (MRS)[3]
  • in the intraintestinal application of GABA in order to increase intraluminal fluid secretion[4]

Biochem/physiol Actions

GABA (γ-Aminobutyric acid) is an amino acid that functions as an inhibitory neurotransmitter in the central nervous system and as a neuromodulator in some peripheral tissues. It binds to the GABAA receptor and inhibits the activity of signal-receiving neurons. The GABAA receptors are found to form a channel in the cell membranes of neurons. Upon binding of GABA, the receptor is activated to allow the passage of negatively charged molecules, such as chloride ions, into the cell. This results in a lowering of the excitability of the cell.[5]
GABAergic neurons involve myorelaxation, anxiolytic treatment, sedation, and anesthetics. GABA can also influence heart rate and blood pressure.[6]
GABA improves plasma concentration, growth hormone, and protein synthesis in the brain. It also plays a role in reducing blood pressure in humans. GABA acts as a potent hypotensive, diuretic, and antidiabetic agent. It also regulates serum lipids levels and the sensation of pain.[1]
[6]
Major inhibitory neurotransmitter in brain; GABAA and GABAB receptor agonist; increases Cl conductance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Fabian-Fine et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 19(1), 298-310 (1998-12-31)
The mechanosensory organs of arachnids receive diverse peripheral inputs. Little is known about the origin, distribution, and function of these chemical synapses, which we examined in lyriform slit sense organ VS-3 of the spider Cupiennius salei. The cuticular slits of
Isabelle Niespodziany et al.
Epilepsia, 61(5), 914-923 (2020-04-17)
The antiepileptic drug candidate, padsevonil, is the first in a novel class of drugs designed to interact with both presynaptic and postsynaptic therapeutic targets: synaptic vesicle 2 proteins and γ-aminobutyric acid type A receptors (GABAA Rs), respectively. Functional aspects of
S J Mihic et al.
Alcohol health and research world, 21(2), 127-131 (1997-01-01)
The neurotransmitter gamma-aminobutyric acid (GABA) inhibits the activity of signal-receiving neurons by interacting with the GABAA receptor on these cells. The GABAA receptor is a channel-forming protein that allows the passage of chloride ions into the cells. Excessive GABAA activation
Radhika Dhakal et al.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology], 43(4), 1230-1241 (2013-09-14)
GABA (γ-aminobutyric acid) is a four carbon non-protein amino acid that is widely distributed in plants, animals and microorganisms. As a metabolic product of plants and microorganisms produced by the decarboxylation of glutamic acid, GABA functions as an inhibitory neurotransmitter
Richard A E Edden et al.
Magnetic resonance in medicine, 68(3), 657-661 (2012-07-11)
Edited magnetic resonance spectroscopy makes possible noninvasive studies of the role of the inhibitory neurotransmitter GABA in the healthy brain and in disease processes. A major limitation of the methodology is coediting of macromolecular signals. Although it has previously been

Articles

Professor Rivnay (Northwestern University, USA) discusses using organic mixed conductors as an alternative to efficiently bridge the ionic world of biology with contemporary microelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service