Merck
  • [Construction of a lentiviral RNA interference system targeting heparanase based on miR30 and its silencing effect].
[Construction of a lentiviral RNA interference system targeting heparanase based on miR30 and its silencing effect].
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences (2013-03-19)
Xiaoyan Liu, Hong Fang, Dingxian Zhu, Yu Zhang
ABSTRACT
To construct a lentiviral RNA interference system targeting heparanase (HPSE) based on miR30 and to test its silencing effect. Three heparanase-shRNA structures were designed based miR30. The targeting fragments were obtained by PCR, then inserted into the vector LV PP-GFP to construct the recombinant lentiviral vector LV PP-GFP/miR-HPSE-shRNA, which was identified by PCR and sequencing. The 293T cells were co-transfect with LV PP-GFP/miR-HPSE-shRNA, pHelper 1.0 vector and pHelper 2.0 vector to produce lentiviruses, with which A375 cells were infected. Real-time fluorescence quantitative PCR and Western blot were performed to evaluate the expression of heparanase RNA and protein. The lentiviral miR30-based RNAi vector targeting heparanase was constructed and confirmed by PCR and sequencing. The results of real-time fluorescence quantitative PCR and Western blot showed that the expression levels of both heparanase mRNA and protein in infected A375 cells were decreased significantly than those in control group. The lentiviral miR30-based RNAi vector targeting heparanase was been constructed successfully, which can be used for further study on RNAi-mediated oncolytic viruses.
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution