Merck
  • Human liver carboxylesterase 1 outperforms alpha-fetoprotein as biomarker to discriminate hepatocellular carcinoma from other liver diseases in Korean patients.
Human liver carboxylesterase 1 outperforms alpha-fetoprotein as biomarker to discriminate hepatocellular carcinoma from other liver diseases in Korean patients.
International journal of cancer (2013-01-16)
Keun Na, Seul-Ki Jeong, Min Jung Lee, Sang Yun Cho, Sun A Kim, Min-Ji Lee, Si Young Song, Hoguen Kim, Kyung Sik Kim, Hyun Woong Lee, Young-Ki Paik
ABSTRACT
Although alpha-fetoprotein (AFP) is currently the major serologic biomarker for hepatocellular carcinoma (HCC), it cannot efficiently distinguish this cancer from other forms of liver disease in early diagnosis due to its low sensitivity. The aim of this study is to compare sensitivity and specificity of human carboxylesterase 1 (hCE1) and AFP biomarker. Antibody-based assays for hCE1 and AFP were used to test both biomarkers with respect to diagnostic efficiency, Youden's index and the area under the curve (AUC) through receiver operating characteristic (ROC) analysis in plasma from 208 patients with HCC (n=57), liver cirrhosis (n=27), chronic hepatitis (n=37), cholangiocarcinoma (n=22), gastric cancer (n=31) and pancreatic cancer (n=34), along with 52 healthy donors (HDs). The levels of hCE1 were significantly higher in patients with HCC than HDs and the other diseases (p<0.005), further verified by AUC values and Youden's index. In the set of HCC versus liver cirrhosis the AUC values were 0.744 (AFP), 0.918 (hCE1) and 0.938 (combination of AFP and hCE1), respectively. These results indicate that hCE1 is not only a more potent and specific marker in distinguishing cancer from liver diseases, in particular cirrhosis, but the combination of hCE1 and AFP shows also synergistic potential for greater sensitivity and specificity in early diagnosis. Therefore the antibody-based hCE1 assay appears to have high diagnostic efficiency for discriminating HCC from other forms of liver disease. It is now feasible to further validate this novel plasma-based biomarker in the large cohort we assembled.
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in mouse NSO cells, ≥95% (SDS-PAGE)
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Sigma-Aldrich
Carboxylesterase 1 isoform b human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Carboxylesterase 1 isoform c human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein