Merck
  • Mass spectrometry proteomics reveals a function for mammalian CALCOCO1 in MTOR-regulated selective autophagy.
Mass spectrometry proteomics reveals a function for mammalian CALCOCO1 in MTOR-regulated selective autophagy.
Autophagy (2020-01-24)
Jonathan A Stefely, Yu Zhang, Elyse C Freiberger, Nicholas W Kwiecien, Hala Elnakat Thomas, Alexander M Davis, Nathaniel D Lowry, Catherine E Vincent, Evgenia Shishkova, Nicholas A Clark, Mario Medvedovic, Joshua J Coon, David J Pagliarini, Carol A Mercer
ABSTRACT
Macroautophagy/autophagy is suppressed by MTOR (mechanistic target of rapamycin kinase) and is an anticancer target under active investigation. Yet, MTOR-regulated autophagy remains incompletely mapped. We used proteomic profiling to identify proteins in the MTOR-autophagy axis. Wild-type (WT) mouse cell lines and cell lines lacking individual autophagy genes (Atg5 or Ulk1/Ulk2) were treated with an MTOR inhibitor to induce autophagy and cultured in media with either glucose or galactose. Mass spectrometry proteome profiling revealed an elevation of known autophagy proteins and candidates for new autophagy components, including CALCOCO1 (calcium binding and coiled-coil domain protein 1). We show that CALCOCO1 physically interacts with MAP1LC3C, a key protein in the machinery of autophagy. Genetic deletion of CALCOCO1 disrupted autophagy of the endoplasmic reticulum (reticulophagy). Together, these results reveal a role for CALCOCO1 in MTOR-regulated selective autophagy. More generally, the resource generated by this work provides a foundation for establishing links between the MTOR-autophagy axis and proteins not previously linked to this pathway. Abbreviations: ATG: autophagy-related; CALCOCO1: calcium binding and coiled-coil domain protein 1; CALCOCO2/NDP52: calcium binding and coiled-coil domain protein 2; CLIR: MAP1LC3C-interacting region; CQ: chloroquine; KO: knockout; LIR: MAP1LC3-interacting region; MAP1LC3/LC3: microtubule associated protein 1 light chain 3; MEF: mouse embryonic fibroblast; MLN: MLN0128 ATP-competitive MTOR kinase inhibitor; MTOR: mechanistic target of rapamycin kinase; reticulophagy: selective autophagy of the endoplasmic reticulum; TAX1BP1/CALCOCO3: TAX1 binding protein 1; ULK: unc 51-like autophagy activating kinase; WT: wild-type.
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Sodium pyrophosphate dibasic, BioUltra, ≥99.0% (T)
Sigma-Aldrich
β-Glycerol phosphate disodium salt pentahydrate, ≥98.0% (NT)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
2-Chloroacetamide, ≥98%
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)