Skip to Content
Merck

Skip To

A2220

ANTI-FLAG® M2 Affinity Gel

purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

Monoclonal ANTI-FLAG® M2, ANTI-FLAG® M2 Affinity Agarose Gel, Anti-ddddk, Anti-dykddddk

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.32
UNSPSC Code:
12352203
Conjugate:
agarose conjugate
Clone:
M2, monoclonal
Application:
IP, affinity chromatography
Citations:
2532
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


conjugate

agarose conjugate

Quality Segment

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

M2, monoclonal

form

buffered aqueous glycerol solution

analyte chemical class(es)

proteins

technique(s)

affinity chromatography: suitable, immunoprecipitation (IP): suitable

matrix

(4% agarose bead; 45-165μm bead size)

isotype

IgG1

capacity

>0.6 mg/mL, resin binding capacity (FLAG-BAP)

shipped in

wet ice

storage temp.

−20°C

General description

Anti-FLAG M2 Affinity gel is a mouse monoclonal antibody that is covalently attached to agarose. The antibody binds FLAG at the N-terminal, Met-N-terminal, C-terminal and internal locations of fusion proteins. Binding is calcium-independent.

Elution - FLAG® peptide, Glycine, pH 3.5, 3x FLAG® peptide
FLAG® peptide, Glycine, pH3.5, 3x FLAG® peptide

Immunogen

DYKDDDDK

Application

Anti-FLAG® M2 affinity gel has been used for western blotting, immunoprecipitation and for the purification of FLAG fusion proteins.

Learn more product details in our FLAG® application portal.

Physical form

Suspension in buffered saline containing azide as preservative and 50% glycerol

Legal Information

ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

FLAG® tag, 3x FLAG®, DYKDDDDK tag

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F1804A9594M8823
description

purified immunoglobulin, buffered aqueous glycerol solution

description

1.0-1.2 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)

description

clone M2, purified immunoglobulin, buffered aqueous solution (Supplied as a solution in 10 mM sodium phosphate)

description

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

conjugate

agarose conjugate

conjugate

unconjugated

conjugate

CY3 conjugate

conjugate

magnetic beads

clone

M2, monoclonal

clone

M2, monoclonal

clone

M2, monoclonal

clone

M2, monoclonal

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

technique(s)

immunoprecipitation (IP): suitable, affinity chromatography: suitable

technique(s)

-

technique(s)

direct immunofluorescence: 10 μg/mL using mammalian cells fixed with methanol:acetone

technique(s)

affinity chromatography: suitable, immunoprecipitation (IP): suitable

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 11 Questions  
  1. How do I equilibrate the resin and how can I clean it for re-use?

    1 answer
    1. Detailed equilibration and regeneration protocols for this product are provided in the Product Information Sheet:
      http://sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/288/183/a2220bul-ms.pdf

      Helpful?

  2. May I ask a question about the product? Does "1mL" mean 1 mL Slurry plus 50% glycerol, which is 2 mL in total? Same as other (e.g., 10 mL = 50% slurry with total 20 mL volume)

    1 answer
    1. The "1mL" indicates that this product is supplied at a volume of 1 milliliter. This ANTI-FLAG M2 affinity gel is supplied as a 50% suspension in 50% glycerol with 10 mM sodium phosphate and 150 mM sodium chloride, pH 7.4, containing 0.02% (w/v) sodium azide (PBS/A).

      Helpful?

  3. What is the bead size for this product?

    1 answer
    1. As stated in the 'PROPERTIES' section of the product page, the bead size range is 45-165 μm.

      Helpful?

  4. Is there a generic agarose bead available for purchase that can be used as a control to test for non-specific binding, in the case that something is binding to the agarose bead itself rather than the tag? If it is not listed on the site, is there a possibility of obtaining generic agarose beads through alternative means? Alternatively, could any information be provided to determine if another provider's generic agarose beads are produced in a similar manner for this specific purpose?

    1 answer
    1. Yes, there are two options available for a generic agarose bead as a control to test for non-specific binding. One option is Product Number. 4B200, Sepharose® 4B, which can be used for sample preclearing and is equivalent to the beads in the FLAG affinity product. The other option is Product Number: A0919, Mouse IgG−Agarose, which can serve as a conjugated antibody bead control.

      Helpful?

  5. Is there a protocol for regeneration?

    1 answer
    1. Please see the link below to the product data-sheet. Instructions state that the column should be regenerated immediately after use by washing with three column volumes of 0.1 M glycine HCI, pH 3.5. The column should be immediately re-equilibrated in TBS until the effluent is at neutral pH:

      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/288/183/a2220bul-ms.pdf

      Helpful?

  6. Is my lysis buffer compatible with Product No. A2220, ANTI-FLAG® M2 Affinity Gel?

    1 answer
    1. The A2220 product information sheet (under Documents, above) contains a reagent compatibility table located on page 6. We do not recommend addition of SDS or reducing agents such as DTT, DTE or 2-mercaptoethanol to the resin. If you have a detergent listed in the table in a higher than recommended concentration, we recommend trying to dilute the sample before applying to the resin.

      Helpful?

  7. How should I elute my protein when using Product No. A2220, ANTI-FLAG® M2 Affinity Gel?

    1 answer
    1. Elution with the peptide is the most gentle method. Acid elution (0.1 M glycine-HCL pH 3.5) is a more stringent method of elution, and should be evaluated for its effect on your protein if it is to be used in downstream applications. Boiling the resin in sample buffer is the most denaturing condition. If this condition is used, the resin cannot be re-used, due to the presence of SDS and/or reducing agents. The elution information can be viewed on A2220 product information sheet (under Documents, above).

      Helpful?

  8. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  9. When using ANTI-FLAG M2 Affinity Gel, Product A2220, should I use a 3X FLAG peptide or a 1X FLAG peptide to elute my protein?

    1 answer
    1. If you have a 3X FLAG-tagged protein, then you will need to use the 3X FLAG peptide.  If you have a 1X FLAG-tagged protein, you can use the 1X FLAG peptide or the 3X FLAG peptide.  We have not noticed a significant  difference in elution efficiency by using a 3X FLAG peptide on a 1X FLAG-tagged protein.

      Helpful?

  10. When using Product No. A2220, ANTI-FLAG® M2 Affinity Gel, I see bands at 20-25 kDa and 50-60 kDa appearing in my Westerns that are not my FLAG®-tagged protein. How can I prevent this?

    1 answer
    1. As a result of the conjugation, there may be some M2 antibody that is not conjugated to the resin, but is associated with the resin and may appear in acid elutions as heavy and light chain when using the anti-mouse IgG conjugated secondary antibody. We recommend an acid wash (0.1 M glycine-HCL pH 3.5) and neutralization of the resin (do not allow the acid wash to sit on the resin longer than 20 minutes) prior to applying the lysate.  Another way to avoid this is to use a directly conjugated FLAG® antibody for detection such as product A8592 ANTI-FLAG® M2 HRP, or the rabbit anti-FLAG® polyclonal antibody, F7425.

      Helpful?

1–10 of 11 Questions  

Reviews

Active Filters

  1. Baton Rouge
    • Review 1
    • Vote 1
    5 out of 5 stars.

    Works well

    This resin works well at purifying FLAG-tagged proteins very specifically if you do the correct amount of washes (5).

    Helpful?

  2. 1 Ratings-only review