Merck
  • Inhibitory effect of novel 5-O-acyl juglones on mammalian DNA polymerase activity, cancer cell growth and inflammatory response.
Inhibitory effect of novel 5-O-acyl juglones on mammalian DNA polymerase activity, cancer cell growth and inflammatory response.
Bioorganic & medicinal chemistry (2011-09-10)
Sayako Maruo, Isoko Kuriyama, Kouji Kuramochi, Kazunori Tsubaki, Hiromi Yoshida, Yoshiyuki Mizushina
ABSTRACT
We previously found that vitamin K(3) (menadione, 2-methyl-1,4-naphthoquinone) inhibits the activity of human mitochondrial DNA polymerase γ (pol γ). In this study, we focused on juglone (5-hydroxy-1,4-naphthoquinone), which is a 1,4-naphthoquinone derivative, and chemically synthesized novel juglones conjugated with C2:0 to C22:6 fatty acid (5-O-acyl juglones). The chemically modified juglones enhanced mammalian pol inhibition and their cytotoxic and anti-inflammatory activities. The juglone conjugated with oleic acid (C18:1-acyl juglone) showed the strongest inhibition of DNA replicative pol α activity and human colon carcinoma (HCT116) cell growth in 10 synthesized 5-O-acyl juglones. C12:0-Acyl juglone was the strongest inhibitor of DNA repair-related pol λ, as well as the strongest suppression of the production of tumor necrosis factor (TNF)-α production induced by lipopolysaccharide (LPS) in the compounds tested. Moreover, this compound caused the greatest reduction in 12-O-tetradecanoylphorbol-13-acetate (TPA)-induced acute inflammation in mouse ears. C12:0- and C18:1-Acyl juglones selectively inhibited the activities of mammalian pol species, but did not influence the activities of other pols and DNA metabolic enzymes tested. These data indicate that the novel 5-O-acyl juglones target anti-cancer and/or anti-inflammatory agents based on mammalian pol inhibition. Moreover, the results suggest that acylation of juglone is an effective chemical modification to improve the anti-cancer and anti-inflammation of vitamin K(3) derivatives, such as juglone.
Product Description
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Linolenic acid, ~70% (GC)
Supelco
Hexanoic acid, analytical standard
Sigma-Aldrich
Hexanoic acid, purum, ≥98.0% (GC)
Supelco
Linoleic acid, analytical standard
Supelco
Linolenic acid, analytical standard
Sigma-Aldrich
Stearic acid 50, tested according to Ph. Eur.
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
Linoleic acid, ≥99%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Oleic acid, Selectophore, ≥99%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Dodecanoic acid, 98%
Supelco
Oleic acid, analytical standard