Skip to Content
Merck
All Photos(1)

Documents

62160

Supelco

Linolenic acid

analytical standard

Synonym(s):

α-Lnn, cis,cis,cis-9,12,15-Octadecatrienoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH=CH)3(CH2)7CO2H
CAS Number:
Molecular Weight:
278.43
Beilstein:
1727693
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.480 (lit.)
n20/D 1.480

bp

230-232 °C/1 mmHg (lit.)

mp

-11 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

functional group

carboxylic acid
oleic acid

storage temp.

−20°C

SMILES string

CC/C=C\C/C=C\C/C=C\CCCCCCCC(O)=O

InChI

1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h3-4,6-7,9-10H,2,5,8,11-17H2,1H3,(H,19,20)/b4-3-,7-6-,10-9-

InChI key

DTOSIQBPPRVQHS-PDBXOOCHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Linolenic acid has been used in the semisynthesis of lipo-alkaloids derived from aconitine, which may act as anti-inflammatory agents.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

An ω-3 fatty acid that serves as a precursor to eicosapentaenoic acid (EPA) but not docosahexaenoic acid. Conversion is greater in women than men, and conversely, β-oxidation metabolism is greater in men than women.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Semisynthesis and pharmacological investigation of lipo-alkaloids prepared from aconitine
Borcsa B, et al.
Fitoterapia, 82(3), 365-368 (2011)
Aliza H Stark et al.
Nutrition reviews, 66(6), 326-332 (2008-06-05)
Consumption of omega 3 fatty acids is known to have health benefits. For many years, the importance of the only member of the omega 3 family considered to be essential, alpha-linolenic acid (ALA), has been overlooked. Current research indicates that
C Mapiye et al.
Meat science, 95(1), 98-109 (2013-05-15)
Yearling steers were fed 70:30 forage:concentrate diets for 205 d, with either grass hay (GH) or red clover silage (RC) as the forage source, and concentrates containing either sunflower-seed (SS) or flaxseed (FS), each providing 5.4% oil to diets. Feeding
Joel A Simon et al.
The American journal of clinical nutrition, 89(5), 1558S-1564S (2009-03-27)
alpha-Linolenic acid (ALA; 18:3n-3) has been associated inconsistently with an increased risk of prostate cancer. Additional studies have become available since the publication of 2 previous meta-analyses. The objective was to review the published data on the relation between ALA
Takayoshi Suzuki et al.
Journal of medicinal chemistry, 51(23), 7640-7644 (2008-11-15)
A weak, nonselective G protein-coupled receptor 120 (GPR120) agonist 10 was found by screening a series of carboxylic acids derived from the peroxisome proliferator-activated receptor gamma (PPARgamma) agonist 3. Modification based on the homology model of GPR120 led to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service