<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
AF
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
Sodium bromide
Synonym(s):
Sodium bromide
Linear Formula:
NaBr
CAS Number:
7647-15-6
Molecular Weight:
102.89
EC Number:
231-599-9
Products
Product Category
salts (9)
Brand
SAFC (1)
Sigma-Aldrich (7)
Supelco (1)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
ACS reagent (2)
Anhydrous (2)
pharmaceutical primary standard (1)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
310506
Sodium bromide,
ACS reagent, ≥99.0%
Expand
View Pricing
220345
Sodium bromide,
ReagentPlus
®
, ≥99%
Expand
View Pricing
229881
Sodium bromide,
≥99.99% trace metals basis
Expand
View Pricing
71329
Sodium bromide,
BioUltra, ≥99.0% (AT)
Expand
View Pricing
746401
Sodium bromide,
anhydrous, free-flowing, Redi-Dri
™
,
ReagentPlus
®
, ≥99%
Expand
View Pricing
1.06363
Sodium bromide,
99.995 Suprapur
®
Expand
View Pricing
1.06360
Sodium bromide,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JP, USP
Expand
View Pricing
S4547
Sodium bromide,
BioXtra, ≥99.0%
Expand
View Pricing
1613597
Sodium bromide,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
793574
Sodium bromide,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥99%
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More