Skip to Content
Merck
All Photos(7)

Key Documents

746401

Sigma-Aldrich

Sodium bromide

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBr
CAS Number:
Molecular Weight:
102.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor pressure

1 mmHg ( 806 °C)

product line

ReagentPlus®
Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

pH

5.4 (20 °C, 50 g/L)

mp

755 °C (lit.)

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Co-catalyst for TEMPO/NaOCl oxidation of alcohols in non-chlorinated solvents.

Towards greener solvents for the bleach oxidation of alcohols catalysed by stable N-oxy radicals

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Cazón et al.
International journal of biological macromolecules, 117, 235-246 (2018-05-25)
The aim of this study was to develop composite films from cellulose, chitosan and polyvinyl alcohol to obtain environmentally friendly materials. Toughness, burst strength, distance to burst and water adsorption properties were measured and analysed as a function of cellulose
Patricia Cazón et al.
Carbohydrate polymers, 216, 72-85 (2019-05-03)
Bacterial cellulose (BC) produced by Komagataeibacter xylinus is a biomaterial with a unique three-dimensional structure. To improve the mechanical properties and reinforce the BC films, they were immersed in polyvinyl alcohol (0-4%) and chitosan (0-1%) baths. Moisture content, mechanical properties
Christian Freese et al.
Particle and fibre toxicology, 11, 68-68 (2014-12-30)
In general the prediction of the toxicity and therapeutic efficacy of engineered nanoparticles in humans is initially determined using in vitro static cell culture assays. However, such test systems may not be sufficient for testing nanoparticles intended for intravenous application.
Ramon Weishaupt et al.
Biomacromolecules, 16(11), 3640-3650 (2015-09-29)
Controlled and efficient immobilization of specific biomolecules is a key technology to introduce new, favorable functions to materials suitable for biomedical applications. Here, we describe an innovative and efficient, two-step methodology for the stable immobilization of various biomolecules, including small
Hiroaki Takeda et al.
Journal of bioscience and bioengineering, 120(4), 476-482 (2015-07-15)
Lipids play important roles in the body and are transported to various tissues via lipoproteins. It is commonly assumed that alteration of lipid levels in lipoproteins leads to dyslipidemia and serious diseases such as coronary artery disease (CAD). However, lipid

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service