Skip to Content
Merck

1613597

USP

Sodium bromide

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
NaBr
CAS Number:
Molecular Weight:
102.89
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor pressure

1 mmHg ( 806 °C)

API family

sodium bromide

manufacturer/tradename

USP

mp

755 °C (lit.)

application(s)

pharmaceutical (small molecule)
pharmaceutical

format

neat

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium bromide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Sodium Bromide Compounded Injection, Veterinary
  • Sodium Bromide Compounded Oral Solution, Veterinary
  • Potassium Bromide Compounded Oral Solution, Veterinary

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Sodium bromide


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class

13 - Non Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library