Skip to Content
Merck
All Photos(3)

Key Documents

229881

Sigma-Aldrich

Sodium bromide

≥99.99% trace metals basis

Synonym(s):

Bromide salt of sodium, Trisodium tribromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBr
CAS Number:
Molecular Weight:
102.89
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050404
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis
form:
crystalline

vapor pressure

1 mmHg ( 806 °C)

Quality Level

Assay

≥99.99% trace metals basis

form

crystalline

impurities

≤100.0 ppm Trace Metal Analysis

mp

755 °C (lit.)

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium bromide is a brominating agent mainly used in organic synthetic reactions as a bromide source.

Application

Sodium bromide can be used as:
  • A catalyst to synthesize tetrahydrobenzo[b]pyrans via three-component cyclocondensation of aryl aldehydes, alkyl nitriles, and dimedone under solvent-free and microwave-assisted reactions.
  • A precursor to synthesize bromomethylated γ-lactones via an electrochemical process.
  • A sodium bromide bath to prepare small Ag nanocubes with sharp edges.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Continuous Synthesis of Monodisperse Ag Nanocubes
Albuquerque, et al.
Crystal Growth & Design, 18, 119-125 (2018)
Electrochemical oxidative bromolactonization of unsaturated carboxylic acids with sodium bromide: Synthesis of bromomethylated ?-lactones
Rabin Kim, et al.
Tetrahedron Letters, 88, 153567-153567 (2022)
Sodium bromide catalysed one-pot synthesis of tetrahydrobenzo [b] pyrans via a three-component cyclocondensation under microwave irradiation and solvent free conditions
Devi I and Bhuyan P J
Tetrahedron Letters, 45, 8625-8627 (2004)
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 8b, 943-943 (2014)
Ye Tang et al.
Biosensors, 10(9) (2020-09-17)
Glucose sensors are potentially useful tools for monitoring the glucose concentration in cell culture medium. Here, we present a new, low-cost, and reproducible sensor based on a cellulose-based material, 2,2,6,6-tetramethylpiperidine-1-oxyl (TEMPO) oxidized-cellulose nanocrystals (CNCs). This novel biocompatible and inert nanomaterial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service