Skip to Content
Merck
All Photos(1)

Key Documents

1612539

USP

Sertraline hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(1S,4S)-4-(3,4-Dichlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17NCl2 · HCl
CAS Number:
Molecular Weight:
342.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sertraline

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

ClC1=C(Cl)C=CC([C@H]2C3=C(C=CC=C3)[C@@H](NC)CC2)=C1.[H]Cl

InChI

1S/C17H17Cl2N.ClH/c1-20-17-9-7-12(13-4-2-3-5-14(13)17)11-6-8-15(18)16(19)10-11;/h2-6,8,10,12,17,20H,7,9H2,1H3;1H/t12-,17-;/m0./s1

InChI key

BLFQGGGGFNSJKA-XHXSRVRCSA-N

Gene Information

human ... SLC6A4(6532)

Looking for similar products? Visit Product Comparison Guide

General description

Sertraline hydrochloride is a SSRI i.e selective serotonin reuptake inhibitor. It is mainly used in treating psychiatric disorders. It also is effective in treating hypotension occurring in disorders like neurocardiogenic syncope and idiopathic orthostatic hypotension which is affected by autonomic dysfunction.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sertraline hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Sertraline Hydrochloride Oral Solution
  • Sertraline Hydrochloride Tablets

Biochem/physiol Actions

Selective serotonin reuptake inhibitor, antidepressant.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of sertraline hydrochloride on dialysis hypotension.
Dheenan S
American Journal of Kidney Diseases, 31(4), 624-630 (1998)
Anton Pottegård et al.
European child & adolescent psychiatry, 23(12), 1211-1218 (2014-02-05)
Our objective was to describe the use of selective serotonin reuptake inhibitors (SSRIs) in the entire Danish population of children and adolescents from 1995 to 2011. Data on filled SSRIs were obtained for all children in Denmark aged 5-17 during
Julia Morrison et al.
International journal of emergency mental health, 16(1), 247-250 (2014-10-28)
Suicide is one of the major causes of morbidity and mortality amongst children and adolescents. In 2004 the Food and Drug Administration (FDA) issued a "black-box" warning for antidepressants in children and adolescents, stating that these drugs may increase suicidality
Nachshon Korem et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(12), 2709-2722 (2014-06-06)
Posttraumatic stress disorder (PTSD) can develop following exposure to a traumatic event. Hence, what we do in the first few hours after trauma exposure may alter the trajectory of PTSD. We examined whether cannabinoids can prevent the effects of a
Melanie L Hedgespeth et al.
Aquatic toxicology (Amsterdam, Netherlands), 151, 84-87 (2014-01-02)
Pharmaceutical residues are increasingly detected in environmental and biological samples, some at levels known to adversely affect non-target organisms; however, less is known of how these organism-level effects relate to the ecology of aquatic systems. Foraging processes may be used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service