Skip to Content
Merck
All Photos(1)

Key Documents

1356756

USP

Lamotrigine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, GI 267119X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7Cl2N5
CAS Number:
Molecular Weight:
256.09
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lamotrigine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1nnc(c(N)n1)-c2cccc(Cl)c2Cl

InChI

1S/C9H7Cl2N5/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16-15-7/h1-3H,(H4,12,13,14,16)

InChI key

PYZRQGJRPPTADH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anticonvulsant.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.
Sales restrictions may apply.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Permeation enhancers on diffusion of lamotrigine drug through cellophane membrane.
Rao, Vinay, et al.
American Journal of Advanced Drug Delivery, 1.4, 606-610 (2013)
Allyson K Friedman et al.
Science (New York, N.Y.), 344(6181), 313-319 (2014-04-20)
Typical therapies try to reverse pathogenic mechanisms. Here, we describe treatment effects achieved by enhancing depression-causing mechanisms in ventral tegmental area (VTA) dopamine (DA) neurons. In a social defeat stress model of depression, depressed (susceptible) mice display hyperactivity of VTA
Mary Ellen Trunko et al.
The International journal of eating disorders, 47(3), 329-334 (2013-12-18)
Some patients with symptoms of binge eating and purging are successfully treated with specific serotonin reuptake inhibitors (SSRIs), but others experience only partial or no benefit. Significant affect dysregulation and poor impulse control may be characteristics that limit responsiveness. We
Jacqueline A French et al.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics, 9(1), 176-184 (2011-12-06)
The efficacy and safety of lamotrigine extended-release tablets (LTG XR) as monotherapy for partial seizures were evaluated using the conversion-to-monotherapy design, and historical data as the control. This methodology was recently approved by the United States Food and Drug Administration
Philip J Wiffen et al.
The Cochrane database of systematic reviews, (2)(2), CD006044-CD006044 (2011-02-18)
This is an update of the original Cochrane review published in Issue 2, 2007. Some antiepileptic medicines have a place in the treatment of neuropathic pain (pain due to nerve damage). This updated review adds five new additional studies looking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service