Merck
  • Lamotrigine use in patients with binge eating and purging, significant affect dysregulation, and poor impulse control.
Lamotrigine use in patients with binge eating and purging, significant affect dysregulation, and poor impulse control.
The International journal of eating disorders (2013-12-18)
Mary Ellen Trunko, Terry A Schwartz, Enrica Marzola, Angela S Klein, Walter H Kaye
ABSTRACT
Some patients with symptoms of binge eating and purging are successfully treated with specific serotonin reuptake inhibitors (SSRIs), but others experience only partial or no benefit. Significant affect dysregulation and poor impulse control may be characteristics that limit responsiveness. We report on the treatment of five patients with bulimia nervosa (BN), anorexia nervosa-binge/purge type (AN-B/P) or eating disorder not otherwise specified (EDNOS), using the anticonvulsant lamotrigine after inadequate response to SSRIs. Following addition of lamotrigine to an antidepressant in four cases, and switch from an antidepressant to lamotrigine in one case, patients experienced substantial improvement in mood reactivity and instability, impulsive drives and behaviors, and eating-disordered symptoms. These findings raise the possibility that lamotrigine, either as monotherapy or as an augmenting agent to antidepressants, may be useful in patients who binge eat and purge, and have significant affect dysregulation with poor impulse control.
Product Description
Supelco
Lamotrigine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Lamotrigine, ≥98%, powder
Lamotrigine for peak identification, European Pharmacopoeia (EP) Reference Standard
Lamotrigine, European Pharmacopoeia (EP) Reference Standard
USP
Lamotrigine, United States Pharmacopeia (USP) Reference Standard
Lamotrigine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Lamotrigine, Pharmaceutical Secondary Standard; Certified Reference Material