Skip to Content
Merck
All Photos(1)

Key Documents

1188800

USP

Diclofenac sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Diclofenac sodium salt, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10Cl2NNaO2
CAS Number:
Molecular Weight:
318.13
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diclofenac

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[O-]C(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl

InChI

1S/C14H11Cl2NO2.Na/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;/h1-7,17H,8H2,(H,18,19);/q;+1/p-1

InChI key

KPHWPUGNDIVLNH-UHFFFAOYSA-M

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Diclofenac sodium is a sodium salt of diclofenac, a non-steroidal anti-inflammatory drug (NSAID). It prevents the synthesis of prostaglandin involved in pain, inflammation, and fever by inhibiting the cyclooxygenase enzyme activity.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diclofenac sodium USP reference standard suitable for use in specified USP compendial quality tests and assays.

Also used to prepare acid stage standard, standard, standard stock, sensitivity, and system suitability solution for the assay, impurity analysis, identification, and performance test by using liquid chromatography coupled with UV detector according to the given below monographs of United States Pharmacopeia (USP): 
  • Diclofenac Sodium
  • Diclofenac Sodium Delayed-Release Tablets  
  • Diclofenac Sodium Extended-Release Tablets
  • Diclofenac Sodium Topical Solution
  • Diclofenac Sodium and Misoprostol Delayed-Release Tablets

Biochem/physiol Actions

Standard NSAID and cyclooxygenase (COX) inhibitor. Major metabolites are 4´-hydroxydiclofenac and 5´-hydroxydiclofenac. Has been used as substrate selective for CYP2C9.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diclofenac Sodium
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 42(4), 1347-1347 (2020)
Adaptation of a color reaction for spectrophotometric determination of diclofenac sodium and piroxicam in pure form and in pharmaceutical formulations
El-Didamony, AM and Amin, AS
Analytical Letters, 37(6), 1151-1162 (2004)
Diclofenac Sodium and Misoprostol Delayed-Release Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(5), 1353-1353 (2023)
Diclofenac Sodium Extended-Release Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(6), 1350-1350 (2016)
Diclofenac Sodium Delayed-Release Tablets
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 40(6), 1348-1348 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service