Skip to Content
Merck
All Photos(1)

Key Documents

1046067

USP

Azithromycin Identity

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Azithromycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H72N2O12
CAS Number:
Molecular Weight:
748.98
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

azithromycin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C38H72N2O12/c1-15-27-38(10,46)31(42)24(6)40(13)19-20(2)17-36(8,45)33(52-35-29(41)26(39(11)12)16-21(3)48-35)22(4)30(23(5)34(44)50-27)51-28-18-37(9,47-14)32(43)25(7)49-28/h20-33,35,41-43,45-46H,15-19H2,1-14H3/t20-,21-,22+,23-,24-,25+,26+,27-,28+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1

InChI key

MQTOSJVFKKJCRP-BICOPXKESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Azithromycin is a subclass of macrolides consisting of 15-membered macrocyclic lactone ring to which sugar moieties are linked.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

It was used as calibration standard at concentrations 3.90, 5.85, 7.80, 9.75 and 11.70 mg/mL during HPLC analysis to determine azithromycin in tablets and capsules.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Development and validation of a micellar liquid chromatographic method with UV detection for determination of azithromycin in tablets and capsules.
Kulikov, A. U., and A. G. Verushkin.
Chromatographia, 60.1-2, 33-38 (2004)
Aspects of degradation kinetics of azithromycin in aqueous solution.
Zhang, Yu, et al.
Chromatographia, 70.1-2, 67-73 (2009)
Sheila K West et al.
The Journal of infectious diseases, 210(1), 65-71 (2014-01-22)
Trachoma, caused by repeated infections with ocular Chlamydia trachomatis, is targeted for elimination using multiple annual rounds of mass drug administration (MDA) in endemic communities. Infection rates do not decline as expected in some communities, leading to concerns about azithromycin
Eric M Mortensen et al.
JAMA, 311(21), 2199-2208 (2014-06-04)
Although clinical practice guidelines recommend combination therapy with macrolides, including azithromycin, as first-line therapy for patients hospitalized with pneumonia, recent research suggests that azithromycin may be associated with increased cardiovascular events. To examine the association of azithromycin use with all-cause
Katharina Krenn et al.
The Journal of thoracic and cardiovascular surgery, 149(4), 1194-1202 (2015-01-18)
Azithromycin has become a standard of care in therapy of bronchiolitis obliterans following lung transplantation. Matrix metalloprotease-9 broncho-alveolar lavage levels increase in airway neutrophilia and bronchiolitis obliterans. Interleukin-17 may play a role in lung allograft rejection, and interleukin-12 is downregulated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service