Skip to Content
Merck

1045600

USP

Azaerythromycin A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

9-Deoxo-9a-aza-9a-homoerythromycin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C37H70N2O12
CAS Number:
Molecular Weight:
734.96
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

azithromycin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N1[C@@H]([C@H]([C@@]([C@H](OC(=O)[C@@H]([C@H]([C@@H]([C@H]([C@@](C[C@H](C1)C)(O)C)O[C@@H]3O[C@@H](C[C@@H]([C@H]3O)N(C)C)C)C)O[C@@H]2O[C@H]([C@@H]([C@@](C2)(OC)C)O)C)C)CC)(O)C)O)C

InChI

1S/C37H70N2O12/c1-14-26-37(10,45)30(41)23(6)38-18-19(2)16-35(8,44)32(51-34-28(40)25(39(11)12)15-20(3)47-34)21(4)29(22(5)33(43)49-26)50-27-17-36(9,46-13)31(42)24(7)48-27/h19-32,34,38,40-42,44-45H,14-18H2,1-13H3/t19-,20-,21+,22-,23-,24+,25+,26-,27+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1

InChI key

HRKNNHYKWGYTEN-HOQMJRDDSA-N

General description

Azaerythromycin is a cladinose containing macrolide antibiotic which is a 15-member ring.

Application

Azaerythromycin A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Azithromycin
  • Azithromycin Capsules
  • Azithromycin for Injection

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Azaerythromycin A


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library