Skip to Content
Merck
All Photos(1)

Key Documents

Y0000306

Azithromycin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Azithromycin dihydrate, 1-Oxa-6-azacyclopentadecan-15-one, 13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-alpha-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10- trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-beta-D-xylohexopyranosyl]oxy]-, [2R-(2R, 3S, 4R, 5R, 8R, 10R, 11R, 12S, 13S, 14R)]- dihydrate, N-Methyl-11-aza-10-deoxo-10-dihydroerythromycin A, CP-62993

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H72N2O12 · 2H2O
CAS Number:
Molecular Weight:
785.02
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

azithromycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C38H72N2O12.2H2O/c1-15-27-38(10,46)31(42)24(6)40(13)19-20(2)17-36(8,45)33(52-35-29(41)26(39(11)12)16-21(3)48-35)22(4)30(23(5)34(44)50-27)51-28-18-37(9,47-14)32(43)25(7)49-28;;/h20-33,35,41-43,45-46H,15-19H2,1-14H3;2*1H2/t20-,21-,22+,23-,24-,25+,26+,27-,28+,29-,30+,31-,32+,33-,35+,36-,37-,38-;;/m1../s1

InChI key

SRMPHJKQVUDLQE-KUJJYQHYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Azithromycin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Azithromycin dihydrate is a macrolide antibiotic with anti-immunomodulatory/anti-inflammatory properties.
Azithromycin dihydrate is a macrolide antibiotic, azalide subclass. It binds to the 50S subunit of the 70S bacterial ribosomes and inhibits RNA-dependent protein synthesis in bacterial cells. Azithromycin also has anti-immunomodulatory/anti-inflammatory properties, which make it useful in treating cystic fibrosis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katharina Krenn et al.
The Journal of thoracic and cardiovascular surgery, 149(4), 1194-1202 (2015-01-18)
Azithromycin has become a standard of care in therapy of bronchiolitis obliterans following lung transplantation. Matrix metalloprotease-9 broncho-alveolar lavage levels increase in airway neutrophilia and bronchiolitis obliterans. Interleukin-17 may play a role in lung allograft rejection, and interleukin-12 is downregulated
Patricia C Valery et al.
The Lancet. Respiratory medicine, 1(8), 610-620 (2014-01-28)
Indigenous children in high-income countries have a heavy burden of bronchiectasis unrelated to cystic fibrosis. We aimed to establish whether long-term azithromycin reduced pulmonary exacerbations in Indigenous children with non-cystic-fibrosis bronchiectasis or chronic suppurative lung disease. Between Nov 12, 2008
Josje Altenburg et al.
JAMA, 309(12), 1251-1259 (2013-03-28)
Macrolide antibiotics have been shown beneficial in cystic fibrosis (CF) and diffuse panbronchiolitis, and earlier findings also suggest a benefit in non-CF bronchiectasis. To determine the efficacy of macrolide maintenance treatment for adults with non-CF bronchiectasis. The BAT (Bronchiectasis and
Meron Haile et al.
The American journal of tropical medicine and hygiene, 89(4), 717-720 (2013-09-05)
Latrine use has been promoted as a component of an integrated strategy for trachoma control. As part of a randomized trial in Ethiopia, 12 communities received a mass azithromycin distribution followed by a latrine promotion intervention. A random sample of
Keith C Meyer et al.
The European respiratory journal, 44(6), 1479-1503 (2014-11-02)
Bronchiolitis obliterans syndrome (BOS) is a major complication of lung transplantation that is associated with poor survival. The International Society for Heart and Lung Transplantation, American Thoracic Society, and European Respiratory Society convened a committee of international experts to describe

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service