Skip to Content
Merck
All Photos(1)

Key Documents

Z226262

Nucleosil® CN HPLC Column

5 μm particle size, L × I.D. 25 cm × 4.6 mm

Synonym(s):

Cyanopropyl HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52
Pricing and availability is not currently available.

Agency

suitable for USP L10

Quality Level

product line

Nucleosil®

feature

endcapped: no

manufacturer/tradename

Nucleosil®

extent of labeling

5% Carbon loading

parameter

400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

350 m2/g

matrix

cyano

matrix active group

cyano phase

particle size

5 μm

pore size

100 Å

separation technique

hydrophilic interaction (HILIC)
normal phase
reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is/are the storage solvent(s)? What should I store it in and clean it with for using in reverse-phase?

    1 answer
    1. Reverse the orientation of the column. Flush for a few minutes with a 20% aqueous solution of the organic used in the analysis to remove buffer salts. Flush for about 30 mins with 100% of the organic used in the analysis. The column can be stored with this organic solution. Before reusing the column, flush again for a few minutes with 20% organic to prevent precipitation of buffer salts.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service