Skip to Content
Merck
All Photos(1)

Key Documents

75746

Sigma-Aldrich

Sodium dodecyl sulfate

≥99.0% (GC), dust-free pellets

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161900
eCl@ss:
39093306
PubChem Substance ID:
NACRES:
NA.25

description

anionic

Quality Level

Assay

≥99.0% (GC)

form

rod

mol wt

average mol wt 18,000

aggregation number

62

loss

≤5% loss on drying

CMC

7-10 (lit.)

mp

204 - 207  °C (399 - 405 °F)
204-207 °C (lit.)

transition temp

cloud point >100 °C(lit.)

solubility

H2O: soluble 0.29 gm in 10ml, clear, colorless
H2O: 0.1 M at 20 °C, clear, colorless
130 g/L at 20 °C (68 °F)

density

1 g/cm3

anion traces

chloride (Cl-): ≤200 mg/kg

cation traces

Ca: ≤25 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤200 mg/kg
Mg: ≤20 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

HLB

40(lit.)

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium dodecyl sulfate (SDS) is an organic sodium salt used as both a detergent and a protein denaturant in biochemical research. Its unique structure includes a long-chain aliphatic group with a negatively charged sulfate, making it an effective agent for denaturing proteins. This property is especially valuable in electrophoresis, a technique used to separate and analyze macromolecules like proteins. SDS, with its negative charge, interacts with proteins and lipids to create micellar structures, aiding in solubilizing and stabilizing these biomolecules.

Application

Anionic detergent
Sodium dodecyl sulfate (SDS) has been used:
  • as a negatively charged surfactant to study its effects on filtration flux and membrane fouling during the microfiltration of the surfactant-stabilized oil emulsion
  • as a component of plant extraction buffer, and transferring buffer for western blot
  • as a component of 5× Laemmli sample buffer (5× LB) and 10× SDS running buffer for sodium dodecyl-sulfate polyacrylamide gel electrophoresis (SDS-PAGE) and immunoblot

Biochem/physiol Actions

Sodium dodecyl sulfate (SDS) effectively binds to the positively charged and the hydrophobic residues of proteins via sulfate groups and alkyl chains. It functions as a vital detergent in the domain of structural biology and protein folding/unfolding studies. SDS can stimulate the creation of α-helices in a protein structure. It has the ability to promote cooperative unfolding in several proteins including bovine serum albumin (BSA), β-lactoglobulin, α-lactalbumin, and hen egg-white lysozyme structures at elevated concentrations. 

Features and Benefits

  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • Tested for trace levels of Anions and Cations
  • High-purity product ideal for biochemical research purposes

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Kamerbeek et al.
Journal of clinical microbiology, 35(4), 907-914 (1997-04-01)
Widespread use of DNA restriction fragment length polymorphism (RFLP) to differentiate strains of Mycobacterium tuberculosis to monitor the transmission of tuberculosis has been hampered by the need to culture this slow-growing organism and by the level of technical sophistication needed
D Wessel et al.
Analytical biochemistry, 138(1), 141-143 (1984-04-01)
A rapid method based on a defined methanol-chloroform-water mixture for the quantitative precipitation of soluble as well as hydrophobic proteins from dilute solutions (e.g., column chromatography effluents) has been developed. The effectiveness of this method is not affected by the
K Fujimoto
Journal of cell science, 108 ( Pt 11), 3443-3449 (1995-11-01)
We propose a new electron microscopic method, the sodium dodecylsulphate (SDS)-digested freeze-fracture replica labeling technique, to study the two-dimensional distribution of integral membrane proteins in cellular membranes. Unfixed tissue slices were frozen with liquid helium, freeze-fractured, and replicated in a
The gross conformation of protein-sodium dodecyl sulfate complexes.
J A Reynolds et al.
The Journal of biological chemistry, 245(19), 5161-5165 (1970-10-10)
Shane Stegeman et al.
Endocrine-related cancer, 22(2), 265-276 (2015-02-12)
The oncogene MDM4, also known as MDMX or HDMX, contributes to cancer susceptibility and progression through its capacity to negatively regulate a range of genes with tumour-suppressive functions. As part of a recent genome-wide association study it was determined that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service