Skip to Content
Merck
All Photos(1)

Key Documents

59357-U

Supelco

Discovery® Cyano (5 µm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Synonym(s):

TM=["Discovery"] HPLC Chromatography

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L10

product line

Discovery®

feature

endcapped

manufacturer/tradename

Discovery®

packaging

1 ea of

extent of labeling

4.5% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

25 cm × 4.6 mm

surface area

200 m2/g

surface coverage

3.5 μmol/m2

impurities

<10 ppm metals

matrix

silica gel, high purity, spherical base material
fully porous particle

matrix active group

cyano phase

particle size

5 μm

pore size

180 Å

operating pH range

2-8

application(s)

food and beverages

separation technique

hydrophilic interaction (HILIC)
normal phase
reversed phase

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Low hydrophobicity for rapid elution of hydrophobic analytes
  • Excellent peak shape and retention of strongly basic analytes
  • Retention of polar analytes
  • Unique selectivity
  • Significantly less retention than C18 (typically requires lower % organic mobile phase)
  • Stable, low-bleed LC-MS separations
  • Compatible with highly aqueous organic phases

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Graciela B Arhancet et al.
Journal of medicinal chemistry, 53(16), 5970-5978 (2010-08-03)
A new 1,4-dihydropyridine 5a, containing a cyano group at the C3 position, was recently reported to possess excellent mineralocorticoid receptor (MR) antagonist in vitro potency and no calcium channel-blocker (CCB) activity. In the present study, we report the structure-activity relationships
Christophe Morisseau et al.
Analytical biochemistry, 414(1), 154-162 (2011-03-05)
The microsomal epoxide hydrolase (mEH) plays a significant role in the metabolism of numerous xenobiotics. In addition, it has a potential role in sexual development and bile acid transport, and it is associated with a number of diseases such as
Srdan Verstovsek et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 14(3), 788-796 (2008-02-05)
The discovery of an activating somatic mutation in codon 617 of the gene encoding the Janus kinase (JAK)-2 (JAK2 V617F) in patients with myeloproliferative disorders has opened new avenues for the development of targeted therapies for these malignancies. However, no
K Umekawa et al.
Japanese journal of pharmacology, 84(1), 7-15 (2000-10-24)
We describe the pharmacological characteristics of SM-19712 (4-chloro-N-[[(4-cyano-3-methyl-1-phenyl-1H-pyrazol-5-yl)amino]carbonyl] benzenesulfonamide, monosodium salt). SM-19712 inhibited endothelin converting enzyme (ECE) solubilized from rat lung microsomes with an IC50 value of 42 nM and, at 10 - 100 microM, had no effect on other
Ye Ding et al.
Chemical communications (Cambridge, England), 47(33), 9495-9497 (2011-07-19)
A novel and facile DDQ-mediated dehydrogenation from natural rigid polycyclic acids or flexible alkyl acids to generate lactones is described. The formation of lactones proceeds by a radical ion mechanism, which has been established by DPPH˙-mediated chemical identification, ESR spectroscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service