Skip to Content
Merck
All Photos(1)

Key Documents

CRM55869

Supelco

Clofentezine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3,6-Bis(2-chlorophenyl)-1,2,4,5-tetrazine, Clofentezine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8Cl2N4
CAS Number:
Molecular Weight:
303.15
Beilstein:
7137128
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

182-185 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

Clc1ccccc1-c2nnc(nn2)-c3ccccc3Cl

InChI

1S/C14H8Cl2N4/c15-11-7-3-1-5-9(11)13-17-19-14(20-18-13)10-6-2-4-8-12(10)16/h1-8H

InChI key

UXADOQPNKNTIHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Clofentezine is a tetrazine group containing insecticide used against phytophagous mites on citrus fruits, vines, cotton crops, and ornamentals.
As per Regulation (EC) No. 1107/2009, repealing directive 91/414/EEC, clofentezine is approved for plant protection use in the European Union (EU). A maximum residue limit (MRL) of 0.5 mg/kg and 0.05 mg/kg is applicable for the presence of clofentezine in fresh/frozen fruits and tree nuts respectively, in accordance with Regulation (EC) No. 2015/846. Clofentezine is to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
  • The clofentezine CRM may also find following uses:
  • Development and validation of a high-performance liquid chromatography-diode-array detection (HPLC-DAD) method following a dispersive solid phase extraction using zirconia-coated silica particles for the analysis of 21 pesticide residues in oil samples
  • Development of a sample preparation method based on magnetic mixed hemimicelles dispersive solid-phase extraction (MMHDSPE) to separate three acaricides from three fruit juice samples
  • Multi-residue determination of 60 pesticide residues in hen eggs using QuEChERS extraction followed by an LC-MS/MS or GC-MS/MS quantification
  • Simultaneous analysis of 302 pesticides and contaminants in ten catfish samples by low-pressure GC-MS/MS and UHPLC-MS/MS
  • GC-MS/MS and LC-MS/MS based determination of 314 pesticide residues in raisin samples for their risk exposure assessment
  • Simultaneous analysis of 300 pesticide residues in human hair samples by GC-MS/MS and LC-MS/MS

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanoencapsulation of clofentezine with enhanced acaricidal activity against the two spotted mite, Tetranychus urticae Koch (Acari: Tetranychidae)
Ahmadi Z, et al.
Toxin Reviews, 40(4), 962-970 (2021)
Clinical and laboratory findings in ten Milwaukee patients with the acquired immunodeficiency syndrome or prodromal syndromes.
P A Turner et al.
Wisconsin medical journal, 84(4), 19-22 (1985-04-01)
Xiaoling Yang et al.
Talanta, 166, 93-100 (2017-02-19)
In this research, a novel method using magnetic mixed hemimicelles dispersive solid-phase extraction (MMHDSPE) based on C16mimBr-coated attapulgite/polyaniline-polypyrrole/Fe3O4 (ATP/PANI-PPY/Fe3O4) nanocomposites were investigated for enrichment and separation of three acaricides in fruit juice. In this method, we combined the simplicity and
Lijun Han et al.
Food chemistry, 319, 126592-126592 (2020-03-21)
A fast, fit-for-purpose, high-throughput method was developed and validated for the analysis of 302 targeted contaminants in catfish muscle. Targeted contaminants included pesticides and metabolites with US regulatory levels, plus other lipophilic pesticides and environmental contaminants: PAHs, PCBs, PBDEs and
J T Christenson et al.
Nuclear medicine communications, 7(7), 531-540 (1986-07-01)
Left hemispheric blood flow was measured in eight adult Australian sheep prior to and, 1, 4, 30 and 60 min after the injection of endotoxin, E. Coli, 3 mg kg-1 bodyweight, using a radioisotope method. The mean left hemispheric blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service