Skip to Content
Merck
All Photos(1)

Key Documents

48541

Supelco

Trichlorofluoromethane

analytical standard

Synonym(s):

CFC-11, Fluorotrichloromethane, Freon® 11

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CF
CAS Number:
Molecular Weight:
137.37
Beilstein:
1732469
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

5.04 (vs air)

vapor pressure

12.85 psi ( 20 °C)
39.17 psi ( 55 °C)

CofA

current certificate can be downloaded

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.3821 (lit.)

bp

23.7 °C (lit.)

mp

−111-−110 °C (lit.)

density

1.494 g/mL at 25 °C (lit.)

application(s)

PFAS testing
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

FC(Cl)(Cl)Cl

InChI

1S/CCl3F/c2-1(3,4)5

InChI key

CYRMSUTZVYGINF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Freon is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Ozone 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Abdennour Bouhroum et al.
International journal of pharmaceutics, 391(1-2), 98-106 (2010-02-27)
The aims of this study were to investigate and characterize the physico-chemical properties of beclomethasone dipropionate (BDP) crystallized from tricholoromonofluoromethane (CFC-11). Physical interactions in a model pressurised metered dose inhaler (pMDI) system and changes in surface energy after size reduction
T Kyrklund et al.
Scandinavian journal of work, environment & health, 14(2), 91-94 (1988-04-01)
Organic solvents are often present as mixtures in various industrial and house-hold products. The adverse effects arising from exposure to these solvents have often been generalized to concern the whole group of solvents. In an examination of the possibility that
The use of vapocoolants for detumescence.
P W Kutarski
British journal of urology, 72(4), 518-518 (1993-10-01)
Daniele Marmolino et al.
Cerebellum (London, England), 8(3), 245-259 (2009-01-24)
Friedreich's ataxia (FRDA) is a neurodegenerative disease due to a pathological expansion of a GAA triplet repeat in the first intron of the FXN gene encoding for the mitochondrial protein frataxin. The expansion is responsible for most cases of FRDA
Bronchodilator aerosol propellant interferes with an infrared photoacoustic spectrophotometer respiratory gas analyzer.
R W Hoskin et al.
Journal of clinical monitoring, 9(1), 65-66 (1993-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service