Skip to Content
Merck
All Photos(1)

Key Documents

502332

Supelco

Benzo[j]fluoranthene solution

certified reference material, 2000 μg/mL in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
MDL number:
UNSPSC Code:
12352200

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-30°C

SMILES string

c21c3c4c(c2ccc5c1cccc5)cccc4ccc3

InChI

1S/C20H12/c1-2-8-15-13(5-1)11-12-17-16-9-3-6-14-7-4-10-18(19(14)16)20(15)17/h1-12H

InChI key

KHNYNFUTFKJLDD-UHFFFAOYSA-N

General description

Benzo[j]fluoranthene is a polycyclic aromatic hydrocarbon, which is carcinogenic in nature and can be generated into the environment via combustion of coal, coal tar and petroleum products.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Comprehensive Guide to the Hazardous Properties of Chemical Substances (2007)
Report on Carcinogens (12th Ed. ) (2011)
Gemma Assante et al.
Applied microbiology and biotechnology, 69(6), 718-721 (2005-06-30)
The binaphthyl derivative cladosporol 3 was supplied from 60 to 200 mg l(-1) to shaken cultures of Cladosporium cf. cladosporioides. Compared to blank, fungal biomass was not affected by adding cladosporol till 100 mg l(-1): it rather increased at higher
J E Rice et al.
Cancer research, 47(23), 6166-6170 (1987-12-01)
The metabolism of benzo[j]fluoranthene (BjF) in vivo in mouse skin was investigated. trans-4,5-Dihydro-4,5-dihydroxybenzo[j]fluoranthene (BjF-4,5-diol) and trans-9,10-dihydro-9,10-dihydroxybenzo[j]fluoranthene (BjF-9,10-diol) have been identified as major metabolites. In addition, 4- and 10-hydroxybenzo[j]fluoranthene and benzo[j]fluoranthen-4,5-dione have been tentatively identified among the metabolites formed in vivo
E J LaVoie et al.
Cancer letters, 70(1-2), 7-14 (1993-06-15)
Exceptional tumorigenic potency was observed with 4-fluorobenzo[j]fuoranthene (4-fluoroB[j]F) relative to benzo[j]fluoranthene (B[j]F) and 10-fluorobenzo[j]fluoranthene (10-fluoroB[j]F) in a mouse skin initiation promotion bioassay. Comparison of the tumorigenic response obtained at total initiating doses of 50, 100, and 1000 nmol firmly established

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service