Skip to Content
Merck
All Photos(1)

Key Documents

48880-U

Supelco

1,2-Dibromoethane solution

certified reference material, 200 μg/mL in methanol

Synonym(s):

Ethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4Br2
CAS Number:
Molecular Weight:
187.86
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-30°C

SMILES string

BrCCBr

InChI

1S/C2H4Br2/c3-1-2-4/h1-2H2

InChI key

PAAZPARNPHGIKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James K Henderson et al.
Journal of contaminant hydrology, 108(1-2), 29-45 (2009-06-19)
An analytical model is used to simulate the effects of partial source removal and plume remediation on ethylene dibromide (EDB) and 1,2-dichloroethane (1,2-DCA) plumes at contaminated underground storage tank (UST) sites. The risk posed by EDB, 1,2-DCA, and commingled gasoline
James K Henderson et al.
Environmental science & technology, 42(3), 864-870 (2008-03-08)
Field evidence from underground storage tank sites where leaded gasoline leaked indicates the lead scavengers 1,2-dibromoethane (ethylene dibromide, or EDB) and 1,2-dichloroethane (1,2-DCA) may be present in groundwater at levels that pose unacceptable risk. These compounds are seldom tested for
Genotoxic effects of 1,2-dibromoethane and 1,2-dichloroethane.
U Rannug
Mutation research, 76(3), 269-295 (1980-11-01)
J A Hoppin et al.
The European respiratory journal, 34(6), 1296-1303 (2009-06-23)
Although specific pesticides have been associated with wheeze in farmers, little is known about pesticides and asthma. Data from 19,704 male farmers in the Agricultural Health Study were used to evaluate lifetime use of 48 pesticides and prevalent adult-onset asthma
Liping Liu et al.
Chemical research in toxicology, 20(1), 155-163 (2007-01-18)
The production of mutations and the reduction in survival of cells treated with alpha,omega-dihaloalkanes is greatly enhanced by the presence of O6-alkylguanine-DNA alkyltransferase (AGT), a DNA repair protein that removes O6-alkylguanine adducts from DNA [Liu, L., Hachey, D. L., Valadez

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service