Skip to Content
Merck
All Photos(1)

Key Documents

03505

Sigma-Aldrich

1,2-Dibromoethane

purum, ≥98.0% (GC)

Synonym(s):

EDB, Ethylene dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCH2CH2Br
CAS Number:
Molecular Weight:
187.86
Beilstein:
605266
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

~6.5 (vs air)

Quality Level

vapor pressure

11.7 mmHg ( 25 °C)

grade

purum

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.539 (lit.)
n20/D 1.539

bp

131-132 °C (lit.)

mp

8-11 °C (lit.)

density

2.18 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

BrCCBr

InChI

1S/C2H4Br2/c3-1-2-4/h1-2H2

InChI key

PAAZPARNPHGIKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dibromoethane (EDB) is a water-soluble dihaloalkane that was widely used as a soil and grain fumigant. This led to the contamination of soil, surface, and groundwater with residual EDB. As it is a potent carcinogen and mutagen, many studies have been conducted to identify chemical reactions for its degradation in the environment.

Application

1,2-Dibromoethane can be used:
  • To generate vinyl bromide in situ, which can undergo a palladium-catalyzed cross-coupling reaction with aryl boronic acids to form functionalized styrene derivatives.
  • As a brominating agent of tertiary carbanions in sterically hindered molecules.
  • As a building block to synthesize 2-arylsulfonyl hydrazones, (±)-horsfiline and 7-oxo-2, 3-dihydro-7H-pyrido [1,2,3-de] [1,4] benzoxazine-6-carboxylic acids.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 1, 2-dibromoethane (EDB) in field soils: Implications for volatile organic compounds
Sawhney BL, et al.
Journal of Environmental Quality, 17(1), 149-152 (1988)
Persistence of 1, 2-dibromoethane in soils: entrapment in intraparticle micropores
Steinberg SM, et al.
Environmental Science & Technology, 21(12), 1201-1208 (1987)
Synthesis and antibacterial activities of substituted 7-oxo-2,3-dihydro-7H-pyrido [1,2,3-de][1,4] benzoxazine-6-carboxylic acids
Hayakawa I, et al.
Chemical & Pharmaceutical Bulletin, 32(12), 4907-4913 (1984)
Reaction products and rates of disappearance of simple bromoalkanes, 1, 2-dibromopropane, and 1, 2-dibromoethane in water
Vogel TM and Reinhard M
Environmental Science & Technology, 20(10), 992-997 (1986)
1,2-Dibromoethane in the synthesis of 2-bromoesters: Bromination vs alkylation.
Ortuno, Rosa M and others
Tetrahedron, 50(32), 9825-9830 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service