Skip to Content
Merck
All Photos(1)

Documents

P76209

Sigma-Aldrich

Pyruvic acid

95%

Synonym(s):

α-Ketopropionic acid, 2-Oxopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCOOH
CAS Number:
Molecular Weight:
88.06
Beilstein:
506211
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021320
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

refractive index

n20/D 1.428 (lit.)

bp

165 °C (lit.)

mp

11-12 °C (lit.)

density

1.267 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(=O)C(O)=O

InChI

1S/C3H4O3/c1-2(4)3(5)6/h1H3,(H,5,6)

InChI key

LCTONWCANYUPML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emilie A Bard-Chapeau et al.
Nature genetics, 46(1), 24-32 (2013-12-10)
The most common risk factor for developing hepatocellular carcinoma (HCC) is chronic infection with hepatitis B virus (HBV). To better understand the evolutionary forces driving HCC, we performed a near-saturating transposon mutagenesis screen in a mouse HBV model of HCC.
Sarah C Atkinson et al.
Plant molecular biology, 81(4-5), 431-446 (2013-01-29)
Lysine is one of the most limiting amino acids in plants and its biosynthesis is carefully regulated through inhibition of the first committed step in the pathway catalyzed by dihydrodipicolinate synthase (DHDPS). This is mediated via a feedback mechanism involving
Ching-An Wu et al.
Biochimica et biophysica acta, 1833(5), 1147-1156 (2013-02-05)
The Warburg effect is known to be crucial for cancer cells to acquire energy. Nutrient deficiencies are an important phenomenon in solid tumors, but the effect on cancer cell metabolism is not yet clear. In this study, we demonstrate that
Hannes Link et al.
Nature biotechnology, 31(4), 357-361 (2013-03-05)
Recent data suggest that the majority of proteins bind specific metabolites and that such interactions are relevant to metabolic and gene regulation. However, there are no methods to systematically identify functional allosteric protein-metabolite interactions. Here we present an experimental and
Robert A Jacobs et al.
American journal of physiology. Endocrinology and metabolism, 304(7), E686-E694 (2013-02-07)
Lactate is an important intermediate metabolite in human bioenergetics and is oxidized in many different tissues including the heart, brain, kidney, adipose tissue, liver, and skeletal muscle. The mechanism(s) explaining the metabolism of lactate in these tissues, however, remains unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service