Skip to Content
Merck
All Photos(1)

Key Documents

442552

Supelco

Dibromoacetonitrile

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2CHCN
CAS Number:
Molecular Weight:
198.84
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥90% (GC)

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.539 (lit.)

bp

67-69 °C/24 mmHg (lit.)

density

2.296 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

BrC(Br)C#N

InChI

1S/C2HBr2N/c3-2(4)1-5/h2H

InChI key

NDSBDLSWTGLNQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibromoacetonitrile belongs to the family of dihaloacetonitrile compounds that are volatile chlorination by-products. They are proven to be carcinogenic and mutagenic to mice.

Application

Dibromoacetonitrile ( Br2CHCN, CAS Number: 3252-43-5) can be utilized as an analytical reference standard for the quantification of the analyte in water samples by chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis of halonitriles in drinking water using solid-phase microextraction and gas chromatography-mass spectrometry.
Kristiana I, et al.
Journal of Chromatography A, 1225(4), 45-54 (2012)
National Toxicology Program technical report series, (544)(544), 1-193 (2010-08-21)
Dibromoacetonitrile occurs in chlorinated drinking water that contains bromine and interacts with organic matter.We studied the effects of dibromoacetonitrile in drinking water on male and female rats and mice to identify potential toxic or cancer-related hazards. We gave drinking water
Simultaneous and high-throughput analysis of iodo-trihalomethanes, haloacetonitriles, and halonitromethanes in drinking water using solid-phase microextraction/gas chromatography-mass spectrometry: an optimization of sample preparation.
Luo Q, et al.
Journal of Chromatography A, 1365(1-2), 45-53 (2014)
Application of different analytical methods for determination of volatile chlorination by-products in drinking water.
Nikolaou AD, et al.
Talanta, 56(4), 717-726 (2002)
Mohamed H Abdel-Wahab et al.
Pharmacological research, 46(3), 287-293 (2002-09-11)
Dibromoacetonitrile (DBAN) is a disinfection by-product following chlorination of drinking water. Epidemiological studies indicate that it might present a potential hazard to human health. DBAN was previously found to induce oxidative stress in rat stomach as manifested by perturbation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service