Skip to Content
Merck
All Photos(1)

Key Documents

T53805

Sigma-Aldrich

Trichloroacetonitrile

98%

Synonym(s):

Trichlorocyanomethane, Tritox

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCN
CAS Number:
Molecular Weight:
144.39
Beilstein:
605572
EC Number:
MDL number:
UNSPSC Code:
12352117
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

58 mmHg ( 20 °C)

Quality Level

Assay

98%

refractive index

n20/D 1.441 (lit.)

bp

83-84 °C (lit.)

density

1.44 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)(Cl)C#N

InChI

1S/C2Cl3N/c3-2(4,5)1-6

InChI key

DRUIESSIVFYOMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in a synthesis of trichloroacetimidates by DBU catalyzed addition of allylic alcohols and the subsequent study of the MOM catalyzed aza-Claisen rearrangement. Also used to prepare bistrichloroacetimidates from diols leading to dihyrooxazines via acid catalyzed cyclization.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

165.2 °F

Flash Point(C)

74 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christophe Rondot et al.
Organic letters, 9(2), 247-250 (2007-01-16)
An efficient synthesis of chiral dihydrooxazines (2) from 1-aryl-2-amino-propane-1,3-diols (1) via the corresponding bistrichloroacetimidate intermediates has been developed. In this transformation, one trichloroacetimidate acts as a leaving group and the other acts as a nucleophile. The cyclization proceeds through an
Tetrahedron, 63, 2123-2123 (2007)
S A Christ et al.
Journal of toxicology and environmental health, 47(3), 233-247 (1996-02-23)
Trichloroacetonitrile (TCAN) is a by-product of the chlorine disinfection of water containing natural organic material. When administered by gavage to pregnant Long-Evans rats in a medium-chain triglyceride vehicle, tricaprylin oil (Tricap), at a volume of 10 ml/kg, TCAN induced fetal
E L Lin et al.
Cancer letters, 62(1), 1-9 (1992-02-14)
Male Fisher 344 rats were administered 1- or 2-[14C]trichloroacetonitrile (TCAN) by oral gavage. DNA was isolated from the liver, kidneys and stomach and several protein fractions (globin, albumin and globulins) were isolated from blood. TCAN binds to both the DNA
Trichloroacetonitrile.
IARC monographs on the evaluation of carcinogenic risks to humans, 71 Pt 3, 1533-1537 (1999-09-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service