Skip to Content
Merck
All Photos(4)

Key Documents

402923

Sigma-Aldrich

Chloroacetic acid

ACS reagent, ≥99.0%

Synonym(s):

Monochloroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2COOH
CAS Number:
Molecular Weight:
94.50
Beilstein:
605438
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

3.26 (vs air)

vapor pressure

0.75 mmHg ( 20 °C)

Assay

≥99.0%

form

solid

impurities

≤0.01% Carbonyl compounds (as acetone)
≤0.01% Carbonyl compunds (other than acetone)
≤0.01% insolubles

ign. residue

<0.02%

bp

189 °C (lit.)

mp

60-63 °C (lit.)

solubility

water: soluble 3170 g/L at 10 °C

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.002%
heavy metals: ≤0.001% (by ICP)

functional group

carboxylic acid
chloro

SMILES string

OC(=O)CCl

InChI

1S/C2H3ClO2/c3-1-2(4)5/h1H2,(H,4,5)

InChI key

FOCAUTSVDIKZOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloroacetic acid (CAA, monochloroacetic acid, MCAA) is a halogenated aliphatic carboxylic acid. Its mutagenic effects have been studied in Rattus norvegicus. The rate of degradation of MCA by photocatalysis using TiO2 at pH3 is reported to be enhanced under the influence of ozone. CAA is an irritant that hydrolyzes proteins resulting in degradation of the cells. This property has been applied in treating plantar warts.

Application

Chloroacetic acid may be used to synthesize the following:
  • Thiocarbamoylthioacetic acid derivatives.
  • 4-Thiazolone derivatives.
  • Triazinothiazolone derivatives.
  • Water soluble carboxymethyl chitosan.

Disclaimer

“The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.”

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

258.8 °F - closed cup

Flash Point(C)

126 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Novel Three-Component Synthesis of Triazinothiazolones.
Holla BS, et al.
Synthetic Communications, 35(3), 333-340 (2005)
Mohammad Faisal Siddiqui et al.
Ecotoxicology and environmental safety, 65(2), 159-164 (2006-05-02)
Chloroacetic acid (CAA) and chlorobenzene (CB) have been evaluated for in vivo mutagenic potential in Rattus norvegicus, employing the following criteria : (i) chromosomal aberrations (CAs) such as breaks, gaps, exchanges, rings, and multiple aberrations and (ii) micronuclei (MN) induction.
Novel thiazolone derivatives of N-aryl-β-alanines.
Stasevych M, et al.
Chemistry of Heterocyclic Compounds, 47(8), 1050-1052 (2011)
Desulfurization of N-Substituted 2-Thioxothiazolidin-4-ones in Aqueous Solutions of Chloroacetic Acid.
Orlinskii MM.
Russ. J. Org. Chem., 34(12), 1805-1806 (1998)
TiO2 photocatalytic oxidation of monochloroacetic acid and pyridine: influence of ozone.
Kopf P, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 136(3), 163-168 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service