Skip to Content
Merck
All Photos(1)

Key Documents

YBD

Sigma-Aldrich

Yeast Brewers

Debittered, inactive

Synonym(s):

Yeast, brewers, Brewers yeast

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.81

biological source

Saccharomyces cerevisiae

Quality Level

form

powder

application(s)

microbiology

storage temp.

2-8°C

SMILES string

OC(=O)CC1c2c3c(ccc2c4c1cccc4)cccc3

InChI

1S/C19H14O2/c20-18(21)11-17-15-8-4-3-7-14(15)16-10-9-12-5-1-2-6-13(12)19(16)17/h1-10,17H,11H2,(H,20,21)

InChI key

GQNBDGXKDJSVGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brewers yeast from the taxonomic point of view is Saccharomyces cerevisiae. S.cerevisiae is a top-fermenting yeast used in ale fermentation. Dried S. cerevisiae can be used in human food, after treating with mild alkali like sodium carbonate to remove the bitter hops. Yeast ferments and improves the sensorial quality of the beer.

Application

Yeast Brewers has been used as a component in the diet for mass rearing of Aedes albopictus for pupal production. It has also been used to induce acute inflammatory pain in rats.

Preparation Note

Dried without regard for enzyme activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brewer's Saccharomyces yeast biomass: characteristics and potential applications
Ferreira IMPLVO, et al.
Trends in Food Science & Technology, 21(2), 77-84 (2010)
Yeasts in Food, 348-352 (2003)
Preclinical pharmacology profile of CS-706, a novel cyclooxygenase-2 selective inhibitor, with potent antinociceptive and anti-inflammatory effects
Ushiyama S, et al.
European Journal of Pharmacology, 578(1), 76-86 (2008)
Fundamentals of Food Biotechnology, 185-185 (1996)
Validation of a new larval rearing unit for Aedes albopictus (Diptera: Culicidae) mass rearing
Balestrino F, et al.
Testing, 9(3), e91914-e91914 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service