Skip to Content
Merck

YSC1

Yeast from Saccharomyces cerevisiae

Type I

Synonym(s):

(Bakers yeast)

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NA.81
UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Yeast from Saccharomyces cerevisiae, Type I

biological source

Saccharomyces cerevisiae

type

Type I

form

powder or solid

application(s)

food and beverages
microbiology

storage temp.

2-8°C

Quality Level

Application

Yeast from Saccharomyces cerevisiae has been used:
  • as a biocatalyst for the anode of yeast microbial fuel cell
  • for the production of ethanol for fuel use
  • to prepare yeast extract

Biochem/physiol Actions

Yeast from Saccharomyces cerevisiae (Baker′s yeast) is used as a protein supplement, energy booster and immune enhancer. It serves as a dough leavening agent in bread production by converting the fermentable sugars to CO2 and ethanol. Baker′s yeast exhibits aroma, anti-molding and nutritive properties. It acts as a cell factory.

Disclaimer

Autolyzes in aqueous buffer at 37 °C.

General description

Yeast from Saccharomyces cerevisiae (Baker′s yeast) is the oldest microbial starter. It is a facultative anaerobe.

Preparation Note

Dried to avoid inactivation of enzymes.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced ethanol production via electrostatically accelerated fermentation of glucose using Saccharomyces cerevisiae
Mathew AS, et al.
Scientific Reports, 5, 15713-15713 (2015)
Rebeccah J Katzenberger et al.
G3 (Bethesda, Md.), 6(12), 4151-4166 (2016-10-19)
Outcomes of traumatic brain injury (TBI) vary because of differences in primary and secondary injuries. Primary injuries occur at the time of a traumatic event, whereas secondary injuries occur later as a result of cellular and molecular events activated in
A microbial biofuel cell with an air-breathing cathode for in vivo glucose sensing applications
Lee JW
Analytical Methods : Advancing Methods and Applications, 7(8), 3324-3326 (2015)
Engineering baker?s yeast: room for improvement
Randez-Gil F, et al.
Trends in biotechnology, 17(6), 237-244 (1999)
Zachariah P G Olufs et al.
Scientific reports, 8(1), 2348-2348 (2018-02-07)
Genetic variability affects the response to numerous xenobiotics but its role in the clinically-observed irregular responses to general anesthetics remains uncertain. To investigate the pharmacogenetics of volatile general anesthetics (VGAs), we developed a Serial Anesthesia Array apparatus to expose multiple

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service